Skip to Content
Merck
All Photos(1)

Key Documents

V800053

Sigma-Aldrich

Ammonium persulfate

LR, ≥98%

Synonym(s):

AP, APS, Ammonium peroxodisulfate, Ammonium peroxydisulfate, PER

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2S2O8
CAS Number:
Molecular Weight:
228.20
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
grade:
LR
form:
crystalline powder

grade

LR

vapor density

7.9 (vs air)

product line

Vetec

Assay

≥98%

form

crystalline powder

pH

1.0-2 (25 °C, 228 g/L)

anion traces

chloride (Cl-): ≤0.002%

cation traces

Fe: ≤0.003%
Mn: ≤0.0002%

SMILES string

N.N.OS(=O)(=O)OOS(O)(=O)=O

InChI

1S/2H3N.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h2*1H3;(H,1,2,3)(H,4,5,6)

InChI key

ROOXNKNUYICQNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for acrylamide gel polymerization.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Momoko Horikoshi et al.
PLoS genetics, 11(7), e1005230-e1005230 (2015-07-02)
Reference panels from the 1000 Genomes (1000G) Project Consortium provide near complete coverage of common and low-frequency genetic variation with minor allele frequency ≥0.5% across European ancestry populations. Within the European Network for Genetic and Genomic Epidemiology (ENGAGE) Consortium, we
Kenji Tanimura et al.
Blood, 125(18), 2835-2844 (2015-03-04)
Antiphospholipid syndrome (APS) is an autoimmune disorder characterized by thrombosis and/or pregnancy complications. β2-glycoprotein I (β2GPI) complexed with phospholipid is recognized as a major target for autoantibodies in APS; however, less than half the patients with clinical manifestations of APS
Daniela C Ferreira et al.
The journal of physical chemistry. B, 115(6), 1368-1375 (2011-01-28)
This paper investigates the structure of the products obtained from the polymerization of aniline with ammonium persulfate in a citrate/phosphate buffer solution at pH 3 by resonance Raman, NMR, FTIR, and UV-vis-NIR spectroscopies. All the spectroscopic data showed that the
Ayrat M Dimiev et al.
ACS nano, 6(9), 7842-7849 (2012-08-14)
Graphite intercalation compounds (GICs) can be considered stacks of individual doped graphene layers. Here we demonstrate a reversible formation of sulfuric acid-based GICs using ammonium persulfate as the chemical oxidizing agent. No covalent chemical oxidation leading to the formation of
Li-Hua Zhao et al.
Cell research, 25(11), 1219-1236 (2015-10-17)
Strigolactones (SLs) are endogenous hormones and exuded signaling molecules in plant responses to low levels of mineral nutrients. Key mediators of the SL signaling pathway in rice include the α/β-fold hydrolase DWARF 14 (D14) and the F-box component DWARF 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service