Skip to Content
Merck
All Photos(1)

Key Documents

V001947

Sigma-Aldrich

Benzyl alcohol

USP, 98.0-100.5%

Synonym(s):

Benzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
878307
EC Number:
MDL number:
UNSPSC Code:
12352002
Assay:
98.0-100.5%
bp:
203-205 °C (lit.)
vapor pressure:
13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

Agency

USP

vapor density

3.7 (vs air)

vapor pressure

13.3 mmHg ( 100 °C)
3.75 mmHg ( 77 °C)

Assay

98.0-100.5%

form

liquid

autoignition temp.

817 °F

refractive index

n20/D 1.539 (lit.)

bp

203-205 °C (lit.)

mp

−16-−13 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

SMILES string

OCc1ccccc1

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

213.8 °F - DIN 51758

Flash Point(C)

101 °C - DIN 51758


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D W Shaw
American journal of contact dermatitis : official journal of the American Contact Dermatitis Society, 10(4), 228-232 (1999-12-14)
A patient with allergic contact dermatitis caused by benzyl alcohol in a hearing aid impression material and in topical medications is described. In addition, the patient had topical and probably systemic corticosteroid allergy. Benzyl alcohol allergy is reviewed. allergic contact
Endea J Curry et al.
Dermatitis : contact, atopic, occupational, drug, 16(4), 203-208 (2006-03-16)
Benzyl alcohol is a weak sensitizer found in a wide array of plants and plant products, foods, cosmetics, and medications. Delayed contact dermatitis reactions as well as immediate, urticarial, and systemic reactions have been reported to occur with its use
L Gáspár et al.
Fogorvosi szemle, 91(5), 143-150 (1998-06-11)
The authors summarised the literature data of benzylalcohol and amyl-m-cresol. In the clinical practice they used Strepsils in the treatment of 22 patients suffered from oral inflammatic diseases and in the 20 prophylactic cases of oral surgical interventions. The preliminary
J Scognamiglio et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50 Suppl 2, S140-S160 (2011-11-01)
A toxicologic and dermatologic review of benzyl alcohol when used as a fragrance ingredient is presented. Benzyl alcohol is a member of the fragrance structural group Aryl Alkyl Alcohols and is a primary alcohol. The AAAs are a structurally diverse
Benzyl alcohol: a covert fragrance.
Sharon E Jacob et al.
Dermatitis : contact, atopic, occupational, drug, 18(4), 232-233 (2007-11-21)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service