Merck
  • Benzyl alcohol allergy: importance of patch testing with personal products.
Benzyl alcohol allergy: importance of patch testing with personal products.
Dermatitis : contact, atopic, occupational, drug (2006-03-16)
Endea J Curry, Erin M Warshaw
ABSTRACT
Benzyl alcohol is a weak sensitizer found in a wide array of plants and plant products, foods, cosmetics, and medications. Delayed contact dermatitis reactions as well as immediate, urticarial, and systemic reactions have been reported to occur with its use in these various formulations. We present the case of a woman with allergic contact dermatitis from benzyl alcohol and provide a brief review of allergy to this chemical.
Product Description
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Supelco
Benzyl alcohol, analytical standard
USP
Benzyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard