Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
temazepam
manufacturer/tradename
USP
drug control
USDEA Schedule IV; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal); Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet, kontrollierte Droge in Deutschland
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
CN1C(=O)C(O)N=C(c2ccccc2)c3cc(Cl)ccc13
InChI
1S/C16H13ClN2O2/c1-19-13-8-7-11(17)9-12(13)14(18-15(20)16(19)21)10-5-3-2-4-6-10/h2-9,15,20H,1H3
InChI key
SEQDDYPDSLOBDC-UHFFFAOYSA-N
Gene Information
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Analysis Note
Other Notes
Legal Information
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.
English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.
Still not finding the right product?
Explore all of our products under Temazepam
signalword
Warning
hcodes
pcodes
Hazard Classifications
Acute Tox. 4 Oral - STOT SE 3
target_organs
Central nervous system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
