Skip to Content
Merck

Skip To

1445653

USP

Moexipril Related Compound F

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-[(S)-(+)-1-(Ethoxycarbonyl)-3-phenylpropyl]-L-alanine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 mg
Check Cart for Availability
€ 1.410,00

About This Item

Linear Formula:
C6H5CH2CH2CH(CO2C2H5)NHCH(CH3)CO2H
CAS Number:
Molecular Weight:
279.33
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
eCl@ss:
25140904

€ 1.410,00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

moexipril

manufacturer/tradename

USP

mp

150-152 °C (lit.)

application(s)

pharmaceutical
pharmaceutical
pharmaceutical (small molecule)

format

neat

SMILES string

CCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(O)=O

InChI

1S/C15H21NO4/c1-3-20-15(19)13(16-11(2)14(17)18)10-9-12-7-5-4-6-8-12/h4-8,11,13,16H,3,9-10H2,1-2H3,(H,17,18)/t11-,13-/m0/s1

InChI key

CEIWXEQZZZHLDM-AAEUAGOBSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Moexipril Related Compound F USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Moexipril Hydrochloride
  • Enalapril Maleate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
472735PHR30531445642
description

United States Pharmacopeia (USP) Reference Standard

description

98%

description

pharmaceutical secondary standard, certified reference material

description

United States Pharmacopeia (USP) Reference Standard

grade

pharmaceutical primary standard

grade

-

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

application(s)

peptide synthesis

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

format

neat

format

-

format

-

format

neat

API family

moexipril

API family

-

API family

moexipril

API family

moexipril

mp

150-152 °C (lit.)

mp

150-152 °C (lit.)

mp

150-152 °C (lit.)

mp

280-285 °C (lit.)


Still not finding the right product?

Explore all of our products under Moexipril Related Compound F


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters