Skip to Content
Merck

Skip To

1445642

USP

Moexipril Related Compound E

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(S)-(−)-6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 mg
Check Cart for Availability
€ 502,00

About This Item

Empirical Formula (Hill Notation):
C12H15NO4 · HCl
CAS Number:
Molecular Weight:
273.71
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

€ 502,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

moexipril

manufacturer/tradename

USP

mp

280-285 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.COc1cc2CN[C@@H](Cc2cc1OC)C(O)=O

InChI

1S/C12H15NO4.ClH/c1-16-10-4-7-3-9(12(14)15)13-6-8(7)5-11(10)17-2;/h4-5,9,13H,3,6H2,1-2H3,(H,14,15);1H/t9-;/m0./s1

InChI key

ROWPWZMWICGKBY-FVGYRXGTSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1445608PHR3052PHR3606
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

certified reference material, pharmaceutical secondary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

pharmaceutical

format

neat

format

neat

format

-

format

-

mp

280-285 °C (lit.)

mp

-

mp

280-285 °C (lit.)

mp

-

API family

moexipril

API family

moexipril

API family

moexipril

API family

-


Still not finding the right product?

Explore all of our products under Moexipril Related Compound E


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters