Skip to Content
Merck

Skip To

1265457

USP

Ethylbenzene

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

NSC 406903, Phenylethane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
3 x 0.5 mL
Check Cart for Availability
€ 407,00

About This Item

Linear Formula:
C6H5C2H5
CAS Number:
Molecular Weight:
106.17
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1901871

€ 407,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor density

3.7 (vs air)

vapor pressure

10 mmHg ( 20 °C), 19 mmHg ( 37.7 °C)

autoignition temp.

810 °F

expl. lim.

6.7 %

manufacturer/tradename

USP

refractive index

n20/D 1.495 (lit.)

bp

136 °C (lit.)

mp

−95 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCc1ccccc1

InChI

1S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

YNQLUTRBYVCPMQ-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ethylbenzene USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Benzaldehyde

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1229030791.09635
grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

analytical standard

grade

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

application(s)

-

format

neat

format

neat

format

neat

format

-

refractive index

n20/D 1.495 (lit.)

refractive index

n20/D 1.495 (lit.)

refractive index

n20/D 1.495

refractive index

-

density

0.867 g/mL at 25 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

density

0.87 g/cm3 at 20 °C


Still not finding the right product?

Explore all of our products under Ethylbenzene


signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - STOT RE 2

target_organs

hearing organs

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters