Skip to Content
Merck
All Photos(1)

Key Documents

PHR1229

Supelco

Ethylbenzene

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Ethylbenzene, NSC 406903, Phenylethane

Sign Into View Organizational & Contract Pricing

Select a Size

3 X 1.2 ML
€131.00

€131.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
3 X 1.2 ML
€131.00

About This Item

Linear Formula:
C6H5C2H5
CAS Number:
Molecular Weight:
106.17
Beilstein:
1901871
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€131.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1265457

vapor density

3.7 (vs air)

vapor pressure

10 mmHg ( 20 °C)
19 mmHg ( 37.7 °C)

CofA

current certificate can be downloaded

autoignition temp.

810 °F

expl. lim.

6.7 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.495 (lit.)

bp

136 °C (lit.)

mp

−95 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CCc1ccccc1

InChI

1S/C8H10/c1-2-8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

YNQLUTRBYVCPMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards..

Application

Ethylbenzene may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations[1][2] by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3557 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - STOT RE 2

Target Organs

hearing organs

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A validated GC?MS method for the determination and quantification of residual solvents in counterfeit tablets and capsules.
Deconinck E, et al.
Journal of Pharmaceutical and Biomedical Analysis, 70(1-2), 64-70 (2012)
Static headspace gas chromatographic method for quantitative determination of residual solvents in pharmaceutical drug substances according to European Pharmacopoeia requirements.
Otero R, et al.
Journal of Chromatography A, 1057(1-2), 193-201 (2004)
E Jindrová et al.
Folia microbiologica, 47(2), 83-93 (2002-06-13)
Several aerobic metabolic pathways for the degradation of benzene, toluene, ethylbenzene and xylene (BTEX), which are provided by two enzymic systems (dioxygenases and monooxygenases), have been identified. The monooxygenase attacks methyl or ethyl substituents of the aromatic ring, which are
Abby A Li et al.
Neurotoxicology, 31(3), 247-258 (2010-02-23)
The potential for neurotoxicological and immunotoxicological effects of ethylbenzene was studied in young adult Crl:CD(SD) rats following 90-day oral (neurotoxicity) or 28-day inhalation (immunotoxicity) exposures. In the neurotoxicity study, ethylbenzene was administered orally via gavage twice daily at 0, 25
W Tang et al.
Toxicology, 144(1-3), 39-50 (2000-04-27)
In the present studies, human exposure to styrene and to ethylbenzene (EB) is assessed on the basis of literature data. Total styrene and total EB exposure result from inhalation and from food intake. Styrene and EB inhaled represent the greatest

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service