2026-05-20T06:30+00:00
bvseo_fps, prod_bvrr, vn_firebird_3.1.50
cp_1, bvpage1
loc_en_GB, 1241550USP, prd, sort_relevancy
co_noreviews, co_noquestions, tv_0, tr_0
Products
Analytical Chemistry
Analytical Chromatography
Analytical Syringes
Analytical Vials
Carbon Adsorbents
Chromatography Standards
GC Columns, Accessories & Adsorbents
HPLC Buffers
HPLC Columns
LPLC Media & Accessories
Purge Traps
Solvents
TLC Plates & Adsorbents
Analytical Reagents
Acids
Analytical Syringes
Analytical Vials
Caustic Alkalis & Bases
Chromatography & Spectroscopy Reagents
Colorimetric & Titrimetric Test Kits
Indicators
Karl Fischer Reagents
Karl Fischer Standards
Metals
pH Test Strips, pH Papers, Readers & Accessories
Salts
Solvents
Spectroquant Photometry Instruments, Kits, & Accessories
Test Strips, Papers & Readers
Titration Reagents
Analytical Sample Preparation
Air Monitoring Devices & Tubes
Laboratory Filter Holders
Millipore<sup>®</sup> Lab Filter Membranes
Millex<sup>®</sup> Syringe Filters
Purge Traps
Solid Phase Extraction (SPE) & QuEChERS
Solid Phase Micro-Extraction (SPME)
Supported Liquid Extraction (SLE) Columns, Resins & Accessories
Lab Safety
Labware Detergents & Cleaners
Safe Handling
Safety Accessories
Spillage Absorption
Photometry & Rapid Chemical Testing
Colorimetric & Titrimetric Test Kits
pH Test Strips, pH Papers, Readers & Accessories
Photometry Instruments, Test Kits & Accessories
Test Strips, Papers & Readers
Turbidimetric Analysis
Reference Materials
Amino Acid, Peptide & Protein Standards
Biomarker & Metabolite Standards
Cannabis Standards
Carbohydrate Standards
Certified Reference Materials
Chromatography Standards
Food Additive Standards
Food & Cosmetic Component Standards
ICP & AAS Standards
Inorganic Elemental Impurity Mix Standards
Ion Chromatography Standards
Karl Fischer Standards
Lipid Standards
Matrix Materials Standards
Microbiology Standards
Organic Pollutant Standards
Pesticide Standards
Petrochemical Standards
Pharma Standards
Pharmaceuticals, Illicit Drugs & Diagnostic Biomarkers
Pharmacopeia & Metrological Institutes Standards
Physical & Chemical Properties Standards
Phytochemical Standards
Vitamin Standards
Cell Culture & Analysis
3D Cell Culture
Hydrogels
Organoids
Scaffolds
Animal Component Free Cell Culture Reagents
Cell Analysis
Cell Counting Equipment
Cell Health Analysis
Cell Imaging Instruments
Cell Isolation Kits
Cell Migration & Invasion Assays
Live Cell Imaging Reagents
Cell Culture Media & Buffers
Bioprocessing Cell Culture Media
Bioprocessing Liquid Cell Culture Media & Buffers
Classical Media & Buffers
Custom GMP Cell Culture Media
Plant Culture Media
Cell Culture Supplements & Reagents
Albumins & Transport Proteins
Amino Acids
Antibiotics
Attachment Factors
Bioprocessing Recombinant Supplements
Carbohydrates
Cell Dissociation Reagents
Cell Freezing Reagents
FBS
Growth Factors & Cytokines
Hormones
Human Serum
(Other) Sera
Plant Culture Reagents
Sera
Supplements for Cell Culture
Vitamins
Cell Lines & Specialty Cell Culture
Bioprocessing Cell Lines
Mammalian Cell Lines
Mouse Embryo Media
Primary Cells
Stem Cells
Sterile Lab Media Filtration & Cultureware
Cell Culture Supplies
Millex<sup>®</sup> Syringe Filters
Millicell<sup>®</sup> Cell Culture Inserts & Plates
Millipore<sup>®</sup> Sterile Filters
MultiScreen<sup>®</sup> Filter Plates
Stericup<sup>®</sup> Vacuum Filters
Chemistry & Biochemicals
Amino Acids, Resins & Reagents for Peptide Synthesis
Biochemicals
Agarose
Amino Acids
Antibiotics
Avanti Research™, A Croda Brand
Biological Buffers
Carbohydrates
Chelating & Reducing Agents
Coenzymes
Detergents
Lipids
Glycoconjugates
High-Purity Biochemicals & Reagents
Metabolism Assay Kits
Metabolite Libraries
Natural Products
Nucleosides & Nucleotides
Vitamins
Building Blocks
Fluorinated Building Blocks
Halogenated Heterocycles
Heterocyclic Building Blocks
Organic Building Blocks
Catalysts
Buchwald Catalysts & Ligands
C-H Activation Catalysts
Chiral Catalysts & Ligands
Cross-Coupling Catalysts
Hydrogenation Catalysts
Inorganic Catalysts
NHC Ligands & Complexes
Olefin Metathesis
Organocatalysts
Phase-Transfer Catalysts
Phosphine Ligands
Photocatalysts
Transition Metal Catalysts
Chemical Biology
Carbohydrates
Chemical Biology Tools
Click Chemistry Reagents
Crosslinkers
Protein Degrader Building Blocks
Chemical Synthesis
Boronic Acids & Derivatives
C-C Bond Forming Reagents
Chiral Auxiliaries
Coupling Reagents & Nucleosides
Grignard Reagents
Halogenation Reagents
Organoaluminum Reagents
Organolithium Reagents
Organosilicon Reagents
Organotin Reagents
Organozinc Reagents
Oxidation Reagents
Protection/Deprotection Reagents
Reducing Agents
Flavors & Fragrances
Formulation
Biomolecule Formulation
Granulated Materials
Liquid Formulation
Protein Stabilizers
Semisolid Formulation
Solid Formulation
Lab Chemicals
Absorbents & Adsorbents
Acids
Caustic Alkalis & Bases
Drying Agents
Filter Aids & Filter Media
Indicators
Metals
Salts
Solvents
Lab Safety
Labware Detergents & Cleaners
Safe Handling
Safety Accessories
Spillage Absorption
Stable Isotopes
Synthesis Enabling Tools
Automated Chemical Synthesis
High Throughput Screening
Photoreactors
Electrosynthesis
Clinical & Diagnostics
Blood Typing
Blood Typing Finished Reagents
Blood Typing Intermediates For Further Manufacturing Use (FFMU)
Cell Marque™
Cell Marque™ Control Slides
Cell Marque™ Detection Systems & Ancillary Reagents
Cell Marque™ Primary Antibodies
Lateral Flow Membranes
ProClin™ Preservatives
Tissue Diagnostics
Bacteriology Stains
Cell Separation Media
Cytology Stains
Dry Dyes & Certified Stains
Hematology Stains
Histology Stains
Tissue Diagnostics Universal Reagents
Filtration
Concentration Filters
Amicon<sup>®</sup> Centrifugals
Centrifugal Filters
Filter Paper
Filtration for Bioprocessing
Buffer Filtration
Clarification Filter Holders & Systems
Filter Housings
Final Sterile Filtration & Filling
Gas & Vent Filters
Integrity Testers
Media Filtration
Prefilters & Bioburden Control
Sterile Filters
Virus Filter Holders & Systems
Virus Filters
Laboratory Filter Holders
Laboratory Filter Membranes
Filter Membranes
Millipore<sup>®</sup> Filter Membranes
Laboratory Syringe Filters
Syringe Filters
Millex<sup>®</sup> Syringe Filters
MultiScreen<sup>®</sup> Filter Plates
Sterile Laboratory Filter Devices
Millipore<sup>®</sup> Sterile Filters
Stericup<sup>®</sup> Vacuum Filters
Vacuum Filters
Greener Alternative Products
Industrial Microbiology
Environmental Monitoring & Aseptic Process Simulation
Hygiene Monitoring
Microbial Culture Media
Microbial Culture Media Raw Materials
Microbial Filtration
Microbial Identification Tests & Biochemical Reagents
Microbiology Standards
Pathogen Screening Kits, Instruments, Disposables
Pyrogen Testing
Quality Indicator Testing Devices
Sterility Testing Consumables, Media & Instruments
Lab Automation
AAW™ Automated Assay Workstation
Labware
Cell Culture & Cryogenics
Cell Culture Supplies
Cryo Storage
Millicell<sup>®</sup> Cell Culture Inserts & Plates
Filtration
Centrifugal Filters
Filter Membranes
Filter Paper
Laboratory Filter Holders
Syringe Filters
Vacuum Filters
Amicon<sup>®</sup> Centrifugals
Millex<sup>®</sup> Syringe Filters
Millipore<sup>®</sup> Filter Membranes
Millipore<sup>®</sup> Sterile Filters
MultiScreen<sup>®</sup> Filter Plates
Stericup<sup>®</sup> Vacuum Filters
Glassware
Lab Equipment
Balances
Centrifugation
Grinders & Homogenizers
Laboratory Stirring & Mixing
Refrigerators & Freezers
Water Purification
Labware Partners
Benchmark Scientific Products
Brand<sup>®</sup> Products
Corning<sup>®</sup> Products
Cytiva™ Products
DWK Products
Eppendorf<sup>®</sup> Products
Excel Scientific Products
Heathrow Scientific<sup>®</sup> Products
LGC Biosearch™ Products
OHAUS<sup>®</sup> Products
Pop-Bio Products
Roche<sup>®</sup> Products
Liquid Handling & Dispensing
Microscope Slides & Chambers
PCR Supplies & Equipment
Plates
Millicell<sup>®</sup> Cell Culture Inserts & Plates
MultiScreen<sup>®</sup> Filter Plates
MultiScreen<sup>®</sup> Solid Bottom Plates
Sample Handling
Grinders & Homogenizers
Laboratory Containers & Storage
Laboratory Labels
Laboratory Sample Collection
Laboratory Stirring & Mixing
Sealing Films, Foils, Tapes
Septa
Materials Science
Biomedical Materials
3D Bioprinting
Biomedical Polymers
Drug Delivery Formulation Kits
Hydrophilic Polymers
Hydrophobic Polymers
Polyethylene Glycol (PEGs & PEOs)
Polymeric Microspheres & Nanoparticles
Polymerization Tools
Electronic Materials
Carbon Nanomaterials
Electronic Chemicals & Etchants
Inorganic & Metallic Nanomaterials
Liquid Crystals
OLED & PLED Materials
OFET & OPV Materials
Photonic & Optical Materials
Printed Electronic Materials
Self-assembly & Contact Printing
Silicone Materials
Specialty & Smart Polymers
Substrates & Prefab Electronic Components
Energy Materials
Battery Materials
Chalcogenides
Fuel Cell Membranes & Materials
High-Purity Metals & Alloys
High-Purity Salts
Oxides & Ceramics
Quantum Dots
Solar Energy Materials
Solution & Vapor Deposition Precursors
Molecular Biology & Functional Genomics
Cloning & Expression
Molecular Cloning & Protein Expression
Transfection
Epigenetics
Gene Editing & Functional Genomics
CRISPR & Gene Editing
Genetic Screening
RNAi Libraries
RNAi Single Clones
Microbiome
NGS Library Preparation
Nucleic Acid Electrophoresis & Hybridization
Nucleic Acid Electrophoresis, Transfer & Quantitation
Nucleic Acid Labeling
Nucleic Acid Purification
gDNA Purification
Plasmid DNA Purification
Reaction Clean-Up
RNA Purification
Oligos, qPCR Probes & Peptides
Custom & Predesigned DNA Oligos and qPCR Probes
Custom & Predesigned RNA Oligos, siRNA Duplexes and Controls
Custom Peptides
PCR & Amplification
Kapa Biosystems PCR Reagents
PCR Reagent & Kits
PCR Supplies & Equipment
Whole Genome Amplification
Roche<sup>®</sup> Life Science Products
mAbs Development & Manufacturing
Antibody Drug Discovery
Lead Selection & Optimization
Preclinical Development
Target Discovery
Characterization & Quality Control
Aggregate Analysis
Intact Mass Analysis
Peptide Mapping
Post-Translational Modification
Titer Determination
Process Development, Manufacturing, Formulation & Final Fill
Downstream Processing (DSP)
Formulation & Final Fill
Upstream Processing (USP)
mRNA Development & Manufacturing
mRNA Analytical & Quality Control
mRNA Development
mRNA Manufacturing & Formulation
mRNA Research
Pharma & Biopharma Manufacturing
Active Pharmaceutical Ingredients
API Processing Chemicals
Folates
Portfolio APIs
Bioprocessing & Formulation Raw Materials
Bioprocessing Liquid Cell Culture Media & Buffers
Buffers & pH Adjusters
Cleaning in Place
Detergents for Biopharma Applications
Downstream Process & Formulation Chemicals
Endonuclease
High-Quality Chemicals for mRNA Manufacturing
Upstream Process Chemicals
Bioprocessing Cell Culture
Bioprocessing Cell Culture Media
Bioprocessing Cell Lines
Bioprocessing Liquid Cell Culture Media & Buffers
Bioprocessing Recombinant Supplements
Bulk Powder Transfer Bags
Cell Retention
Custom GMP Cell Culture Media
Media Filtration
Viral Vector Upstream Platforms
Bioprocessing Systems
Chromatography Columns for Bioprocessing
Chromatography Systems
Clarification Filter Holders & Systems
Integrity Testers
Single-Use Mixers
Stainless Steel Connectors, Valves & Mixers
Tangential Flow Filtration Holders & Systems
Virus Filter Holders & Systems
Buffer & Media Prep
Buffers & pH Adjusters
Buffer Filtration
Media Filtration
Single-Use Mixers
Chromatography
Affinity Chromatography Resins
Chromatography Columns for Bioprocessing
Chromatography Systems
Ion Exchange Chromatography Resins
Membrane Chromatography
Reverse Phase/Normal Phase Chromatography Resins
Size Exclusion Chromatography Resins
Clarification & Harvest
Clarification Depth Filters & Pretreatment Solutions
Clarification Filter Holders & Systems
Clarification Prefilters
Fluid Management
Connection/Disconnection for Bioprocessing
Fluid Transport & Storage
Single-Use Assemblies
Single-Use Mixers
Stainless Steel Connectors, Valves & Mixers
Formulation
Activated & Functionalized PEGs
Biomolecule Formulation
Granulated Materials
Injectables & Parenteral Formulations
Liquid Formulation
Protein Stabilizers
Semisolid Formulation
Solid Formulation
Synthetic Lipids
Viscosity Reduction Platform
Sterile & Aseptic Sampling Solutions
Sterile Process Sampling
Sampling with MAST<sup>®</sup> Autosampling Solution
Sterile Filtration & Bioburden Control
Buffer Filtration
Filter Housings
Final Sterile Filtration & Filling
Gas & Vent Filters
Integrity Testers
Media Filtration
Prefilters & Bioburden Control
Sterile Filters
Ultrafiltration & Diafiltration
Tangential Flow Filtration Devices
Tangential Flow Filtration Holders & Systems
Viral Clearance
Integrity Testers
Virus Filter Holders & Systems
Virus Filters
Virus Inactivation
Protein Biology
Antibodies
Enhanced Validation Antibodies
Flow Cytometry Antibodies
Prestige Antibodies<sup>®</sup>
Primary Antibodies
Secondary Antibodies
Small Pack Antibodies
ZooMAb<sup>®</sup> Recombinant Antibodies
Bioactive Small Molecules
Structural Genomics Consortium
Duolink<sup>®</sup> PLA - Protein Interaction Technology
Immunoassay Platform Solutions
ELISA Kits
MILLIPLEX<sup>®</sup> Kits
SMC<sup>™</sup> Technology
Protein Electrophoresis & Western Blotting
Electrophoresis & Transfer Equipment
Transfer Membranes & Kits
Immunodetection Reagents & Substrates
Membrane Immunodetection Devices
Protein Electrophoresis Gels & Buffers
Protein Electrophoresis Gel Stains
Protein Markers
Transfer Buffers & Reagents
Proteins & Enzymes
Albumins & Transport Proteins
Coenzymes
Detection Substrates & Enzymes
Drug Analysis Enzymes
Glycobiology Profiling Tools
Nucleases
Proteases
Tissue Dissociation
Protein Mass Spectrometry
Protein Purification Chromatography
Antibody Protein Purification
Biotinylated Protein Purification
FLAG<sup>®</sup> Purification
Recombinant/Fusion Tag Protein Purification
Protein Sample Prep
Amicon<sup>®</sup> Centrifugals
Antibody Protein Purification
Biotinylated Protein Purification
FLAG<sup>®</sup> Purification
Inhibitor Cocktails
Laboratory Concentration Devices
Laboratory Dialysis & Diafiltration Devices
Recombinant/Fusion Tag Protein Purification
Subcellular Fractionation Reagents
Whole Cell Lysis Reagents & Enhancers
Water Purification
Milli-Q<sup>®</sup> Benchtop Water Purification Systems
Milli-Q<sup>®</sup> High-Flow Water Purification Systems
Milli-Q<sup>®</sup> Clinical Water Purification Systems
Milli-Q<sup>®</sup> Consumables
Milli-Q<sup>®</sup> Accessories
Applications
Analytical Chemistry
Atomic Spectroscopy
Calibration, Qualification, Validation
Gas Chromatography (GC)
Liquid Chromatography
Large Molecule HPLC
Low Pressure Liquid Chromatography
Small Molecule HPLC
Mass Spectrometry (MS)
Nuclear Magnetic Resonance (NMR)
Photometry & Reflectometry
Sample Preparation
Sample Filtration
Sample Purification
QuEChERS Sample Preparation
Solid Phase Extraction
Solid Phase Microextraction
Thin Layer Chromatography (TLC)
Titration & Karl Fischer
Water for Analytical Chemistry
Wet Chemical Analysis
Cell Culture & Analysis
Cell Analysis
Cell-based Assays
Cell Counting & Health analysis
Imaging Analysis & Live Cell Imaging
Cell Culture by Cell Type
Mammalian Cell Culture
Microbial Cell Culture
Plant Tissue Culture
Primary Cell Culture
Stem Cell Culture
Cell Culture by Technique
3D Cell Culture
Cell Culture Media Preparation
Cell Growth & Maintenance
Transfection & Gene Editing
Cell Culture for Manufacturing
Cell Culture Troubleshooting
Chemistry & Synthesis
ADC & Bioconjugation
Lead Discovery
Peptide Synthesis
Protein Degradation
Reaction Design & Optimization
Synthetic Methods
C-H Functionalization
Cross Coupling
Fluorination
Metathesis
Organic Reaction Toolbox
Photocatalysis
Clinical & Diagnostics
Bacteriology
Blood Typing
Clinical & Forensics Testing
Cytology & Cytodiagnosis
Diagnostic Immunohistochemistry
Hematology
Histology/Histopathology
IVD Manufacturing
Water for Clinical Diagnostics
Environmental & Cannabis Testing
Air Sampling & Monitoring
Cannabis Testing
Cannabinoids & Potency
Pesticides & Mycotoxins
Terpenes
Drinking Water Testing
Soil, Solid Waste, & Groundwater Testing
Waste Water & Process Water Testing
Food & Beverage Testing & Manufacturing
Chemical Analysis
Flavor & Fragrance Formulation
Microbiological Analysis
Regulatory Compliance
Genomics
Advanced Gene Editing
Cloning & Expression
DNA & RNA Purification
Epigenetics
Functional Genomics Screening
Gene Expression & Silencing
Next Gen Sequencing
Nucleic Acid Gel Electrophoresis
Nucleic Acid Labeling & Detection
PCR
qPCR
Sequencing
Materials Science & Engineering
3D Bioprinting
3D Printing
Batteries, Supercapacitors, Fuel Cells
Bioelectronics
Biosensors & Bioimaging
Chemical Vapor Deposition
Contact Lens & Dental Manufacturing
Drug Delivery
Electron Microscopy
Microelectronics & Nanoelectronics
Nanoparticle & Microparticle Synthesis
Organic Electronics
Photovoltaics & Solar Cells
Polymer Synthesis
Solid State Synthesis
Tissue Engineering
Microbiological Testing
Bioburden Testing
Culture Media Preparation
Media Fill (APS)
Microbial Water Testing
Mycoplasma Testing
Pathogen & Spoilage Testing
Pyrogen Testing
Sterility Testing
Surface & Personnel Testing
Viable Air Monitoring
mAbs Development & Manufacturing
Antibody Drug Discovery
Lead Selection & Optimization
Preclinical Development
Target Discovery
Characterization & Quality Control
Aggregate Analysis
Intact Mass Analysis
Peptide Mapping
Post-Translational Modification
Titer Determination
Process Development, Manufacturing, Formulation & Final Fill
Downstream Processing (DSP)
Formulation & Final Fill
Upstream Processing (USP)
mRNA Development & Manufacturing
mRNA Analytical & Quality Control
mRNA Development
mRNA Manufacturing & Formulation
mRNA Research
Pharma & Biopharma Manufacturing
Antibody Drug Conjugate Manufacturing
Conjugation
Purification
Biopharma 4.0
Bioprocessing Automation & Control Software
Bioprocessing Data Analytics Software
Closed Processing for Biomanufacturing
Intensified & Continuous Upstream (USP) Processing
Intensified & Continuous Downstream (DSP) Processing
Process Analytical Technology (PAT)
Biotech Resource Hub
Discovery
Pre-clinical
Phase I-II
Phase III and Manufacturing
Regulatory
Startup Programs
Blood Plasma Fractionation
Cell Therapy Manufacturing
Classical Pharma Manufacturing
Final Fill Strategies
Liquid Formulation Strategies
Solid Formulation Strategies
Sterile Filtration Strategies
Gene Therapy Manufacturing
Viral Vector Upstream Processing
Viral Vector Downstream Processing
Viral Vector Formulation & Final Fill
Viral Vector Characterization & Biosafety Testing
Viral Vector Contract Development & Manufacturing
Pharma Analysis & QC
Biologics & Biosimilars Characterization
Compendial Testing & Regulatory Guidance
Small Molecules Analysis & Quality Control (QC)
Sterile & Aseptic Sampling
Vaccine Manufacturing
Cell-Based Vaccine Manufacturing
mRNA Vaccine Manufacturing
pDNA Vaccine Manufacturing
Protein Subunit Vaccine Manufacturing
Viral-like Particles Vaccine Manufacturing
Viral Vector Vaccine Manufacturing
Protein Biology
ELISA
Enzyme Activity Assays
Flow Cytometry
Gel Electrophoresis
Immunohistochemistry
Interrogation of Protein Pathways
Lysis & Protein Extraction
Protein & Nucleic Acid Interactions
Protein Concentration & Buffer Exchange
Protein Expression
Protein Labeling & Modification
Protein Pulldown
Protein Purification
Protein Quantitation
Protein Structural Analysis
Western Blotting
Protein Mass Spectrometry
Research & Disease Areas
Cancer Research
Cell & Developmental Biology Research
Cell Signaling
Glycobiology
Metabolomics Research
Neuroscience Research
Pharmacology & Drug Discovery Research
Water Purification
Water for Analytical Chemistry
Water for Cell & Molecular Biology
Water for Clinical Diagnostics
Understanding Lab Water
Services
Contract Manufacturing
ADC & Bioconjugation
Diagnostic OEM & CMO Services
mRNA & LNP
Lipids CDMO Services
Lipid Nanoparticle CDMO Service
Small Molecule
Activated Polyethylene Glycol (PEGs)
APIs
Highly Potent APIs
Viral Vector
AAV Production
Gene Therapy Process Development
Lentiviral Vectors
Contract Testing
Analytical Development
ADC Product Characterization
Analytical Development and Characterization for Adeno-Associated Virus (AAV)
mAb Product Characterization
mRNA Analytical Development
Bulk Harvest Release Testing
Cell & Gene Therapy Bulk Harvest Release Testing
mAbs Bulk Harvest Release Testing
Vaccine Harvest Release Testing
Cell & Virus Bank Manufacturing & Testing
Biorepository & Storage
Cell Bank Manufacturing
Cell Line Characterization
Genetic Stability Testing
Virus Bank Manufacturing
Final Product Release Testing
Cell & Gene Therapy Final Product Release Testing
mAbs Final Product Release Testing
Rapid Molecular Detection
Blazar™ Rapid Molecular Detection
Next Generation Sequencing
Raw Material Testing
Stability Testing Services
Viral Clearance Testing
Provise<sup>®</sup> Clearance Services
Standard & Hybrid Clearance Services
Custom Products
Bulk Powder Transfer Bags
CADI: Custom Assay Development & Innovation
Cell Culture Media Stability & Testing Services
Cell Design Studio™ Cell Engineering Services
CHOZN<sup>®</sup> Cell Line Engineering and Development
Contract ZooMAb<sup>®</sup> Antibody Development Services
Critical Raw Materials
Custom Diagnostic Antibody Development & Manufacturing Services
Custom GC Columns
Custom GMP Cell Culture Media
Custom HPLC Columns
Custom Media Development Services
Custom Nucleic Acids
Custom SPE Tubes & 96-Well Plates
Custom SPME Fibers
Custom Test Strips & Papers
Diagnostic OEM & CMO Services
Eshmuno<sup>®</sup> Fit Custom Affinity Resin Development Services
Production-Scale Materials & Custom Services
Large-Volume Products Supply-Chain
Scalable Manufacturing & Custom Packaging
Reference Materials
Small Molecule Library Design (Aldrich Market Select)
Stable Isotope Custom & GMP Products
Digital Solutions for Life Science
AIDDISON™ AI-Powered Drug Discovery Software
Belysa<sup>®</sup> Immunoassay Curve Fitting Software
Bioprocessing Automation and Control Software
Bioprocessing Data Analytics Software
Catalexis Catalysts Screening Platform
Chemistwin<sup>®</sup> Digital Reference Materials
DEL Extended Digital Service
DOZN™ Tool
mConfig™ Cell Culture Media and Chemicals Configurator
M-Trace<sup>®</sup> Electronic Test Record Software
Millicell<sup>®</sup> Cloud Services
MQuant<sup>®</sup> StripScan
MyMilli-Q<sup>™</sup> Digital Services
SYNTHIA<sup>™</sup> Retrosynthesis Software
Val@M™ Application for Validation Activites
IVD Development & Manufacturing
Custom Diagnostic Antibody Development & Manufacturing Services
Diagnostic OEM & CMO Services
Lateral Flow Assay Development Services
Product Services
Application Services for Solid Dose Formulations
Calibration & Verification
Milli-Q<sup>®</sup> Calibration & Verification
Microbiology Testing Systems
Elevate Program
eMERGE™ Program
Emprove<sup>®</sup> Program
Emprove<sup>®</sup> Cell Culture Media
Emprove<sup>®</sup> Chemicals Portfolio
Emprove<sup>®</sup> Chromatography Portfolio
Emprove<sup>®</sup> Filter and Single-Use Component Portfolio
Emprove<sup>®</sup> Suite
Maintenance & Service Plans
Microbiology Testing Systems
Milli-Q<sup>®</sup> Maintenance & Service Plans
Spectroquant<sup>®</sup> Photometer Services
System Service Reliance Plans
Milli-Q<sup>®</sup> Services & Support
Maintenance & Service Plans
Qualification & Validation
Calibration & Verification
Additional Services
MyMilli-Q™ Digital Services
mPredict™ Co-Crystal Prediction Service
Single-Use & Filter Validation Services
Bacterial Retention Testing
Chemical Compatibility Testing
Extractables & Leachables
Filter Integrity Testing
Validation Service Levels
Validation Services Consultancy
Systems & Equipment Installation
Bioprocessing Systems Equipment Installation
Bioprocessing System Training & Specialized Services
Microbiology Testing Systems
Systems Qualification
Bioprocessing Systems Qualification
Food & Beverages QC Systems Qualification
Milli-Q<sup>®</sup> Qualification & Validation
Pharma QC Systems Qualification
Validation Services
Food & Beverage Microbiology Method Validation
Pharma Microbiology Method Development & Validation
Val@M™ Application for Validation Activites
Support
Inventory Management
M Lab™ Collaboration Centers
Programs
Recycling
Biopharma Recycling Program
Greener Cooler
Polystyrene Cooler Recycling
Training & Education
Resources
Documents
Certificates of Analysis (COA)
Safety Data Sheets (SDS)
Certificates of Quality (COQ)
Certificates of Origin (COO)
Learning Center
Support
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars