Skip to Content
Merck

Skip To

1235398

USP

Enanthic Acid

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Heptanoic acid, Enanthic acid, Oenanthic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
€ 879,00

About This Item

Linear Formula:
CH3(CH2)5COOH
CAS Number:
Molecular Weight:
130.18
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
1744723
MDL number:

€ 879,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor density

4.5 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

API family

enanthic acid

expl. lim.

10.1 %

manufacturer/tradename

USP

refractive index

n20/D 1.4221 (lit.)

bp

223 °C (lit.)

mp

−10.5 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCCC(O)=O

InChI

1S/C7H14O2/c1-2-3-4-5-6-7(8)9/h2-6H2,1H3,(H,8,9)

InChI key

MNWFXJYAOYHMED-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Enanthic Acid USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
43858W33481275190
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

analytical standard

grade

FG, Halal, Kosher, natural

grade

-

application(s)

pharmaceutical (small molecule)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

flavors and fragrances

application(s)

-

format

neat

format

neat

format

-

format

-

density

0.918 g/mL at 25 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

API family

enanthic acid

API family

-

API family

-

API family

-


Still not finding the right product?

Explore all of our products under Enanthic Acid


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters