Skip to Content
Merck
All Photos(1)

Key Documents

43858

Supelco

Heptanoic acid

analytical standard

Synonym(s):

Enanthic acid, Oenanthic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€71.00
5 ML
€310.00

€71.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
1 ML
€71.00
5 ML
€310.00

About This Item

Linear Formula:
CH3(CH2)5COOH
CAS Number:
Molecular Weight:
130.18
Beilstein:
1744723
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€71.00


Estimated to ship on02 June 2025


Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

4.5 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

expl. lim.

10.1 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

refractive index

n20/D 1.4221 (lit.)

bp

223 °C (lit.)

mp

−10.5 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCC(O)=O

InChI

1S/C7H14O2/c1-2-3-4-5-6-7(8)9/h2-6H2,1H3,(H,8,9)

InChI key

MNWFXJYAOYHMED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paolo Lorenzon et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(1), 146-160 (2015-01-09)
The type of neuronal activity required for circuit development is a matter of significant debate. We addressed this issue by analyzing the topographic organization of the olfactory bulb in transgenic mice engineered to have very little afferent spontaneous activity due
Tamar L Greaves et al.
Physical chemistry chemical physics : PCCP, 17(4), 2357-2365 (2014-12-06)
A high-throughput approach was developed in order to prepare and dry a series of protic ionic liquids (PILs) from 48 Brønsted acid-base combinations. Many combinations comprised an alkyl carboxylic acid paired with an alkyl amine. Visual screens were developed to
Pingxi Xu et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(46), 16592-16597 (2014-10-29)
Insect repellents are important prophylactic tools for travelers and populations living in endemic areas of malaria, dengue, encephalitis, and other vector-borne diseases. DEET (N,N-diethyl-3-methylbenzamide) is a 6-decade-old synthetic repellent, which is still considered the gold standard of mosquito repellents. Mosquitoes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service