Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 mg | Check Cart for Availability | € 1.340,00 |
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
tenofovir
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
ClCOC(=O)OC(C)C
InChI
1S/C5H9ClO3/c1-4(2)9-5(7)8-3-6/h4H,3H2,1-2H3
InChI key
JHYNXXBAHWPABC-UHFFFAOYSA-N
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| format neat | format neat | format neat | format neat |
| API family tenofovir | API family propylene carbonate | API family isopropyl acetate | API family amantadine |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. - | storage temp. 2-8°C |
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.





