Skip to Content
Merck
All Photos(1)

Key Documents

SAAR4971590

Phenol Red

pH indicator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116100

product name

Phenol Red,

packaging

poly bottle of 20 × 25 g (SAAR4971590CB)

storage temp.

2-30°C

Application

For laboratory use only

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service