Skip to Content
Merck
All Photos(3)

Key Documents

MMSANSYFU

Millipore

MILLIFLEX OASIS® flow sanitization kit

for use with Milliflex Oasis® pumps

Sign Into View Organizational & Contract Pricing

Select a Size

24 UNITS
€274.00

€274.00


Check Cart for Availability

Request more information

Select a Size

Change View
24 UNITS
€274.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.79

€274.00


Check Cart for Availability

Request more information

packaging

pack of 24 ea

Quality Level

manufacturer/tradename

Milliflex Oasis®

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with Milliflex Oasis® pumps

shipped in

ambient

Related Categories

General description

MILLIFLEX OASIS® consumables for internal flow sanitization kit enables to sanitize the complete pump at once. The components of the kit include a sanitization funnel assembly and syringe. This sanitization kit is a walk-away system to clean efficiently the interior of the MILLIFLEX OASIS® pump.

Application

MILLIFLEX OASIS® consumables for internal flow sanitization kit enables complete sanitization and finds its application in efficient bioburden testing.

Legal Information

MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Milliflex Oasis® system monitors bioburden in various samples, increasing lab productivity.

Milliflex® Rapid System 2.0 accelerates microbial detection in quality control labs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service