Skip to Content
Merck
All Photos(1)

Key Documents

PHL89712

Hypaconitine

phyproof® Reference Substance

Synonym(s):

16,16-trimethoxy-4-(methoxymethyl)-20-methyl-,8-acetate benzoate,(1-α,6-α,14-α,15-α,16-β)-aconitane-8,13,14,15-tetraol

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€492.00

€492.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€492.00

About This Item

Empirical Formula (Hill Notation):
C33H45NO10
CAS Number:
Molecular Weight:
615.71
UNSPSC Code:
41116107
NACRES:
NA.24

€492.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

powder

manufacturer/tradename

PhytoLab

storage temp.

2-8°C

SMILES string

O=C(C1=CC=CC=C1)O[C@H]2[C@@]3(O)[C@@H](OC)[C@H](O)[C@]4(OC(C)=O)C5[C@H](OC)C6C([C@@H](OC)CC[C@]6(CN7C)COC)(C75)[C@H](C3)C42

InChI

1S/C33H45NO10/c1-17(35)44-33-21-19(14-31(38,28(42-6)26(33)36)27(21)43-29(37)18-10-8-7-9-11-18)32-20(40-4)12-13-30(16-39-3)15-34(2)25(32)22(33)23(41-5)24(30)32/h7-11,19-28,36,38H,12-16H2,1-6H3/t19-,20+,21?,22?,23+,24?,25?,26+,27-,28+,30+,31-,32?,33-/m1/s1

InChI key

FIDOCHXHMJHKRW-XCIBRDTCSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service