Skip to Content
Merck
All Photos(1)

Key Documents

PHL85771

Securinine

phyproof® Reference Substance

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€288.00

€288.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€288.00

About This Item

Empirical Formula (Hill Notation):
C13H15NO2
CAS Number:
Molecular Weight:
217.26
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€288.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

2-8°C

SMILES string

O=C1O[C@@]23C[C@@H](C=CC2=C1)N4CCCC[C@H]34

InChI

1S/C13H15NO2/c15-12-7-9-4-5-10-8-13(9,16-12)11-3-1-2-6-14(10)11/h4-5,7,10-11H,1-3,6,8H2/t10-,11-,13+/m1/s1

InChI key

SWZMSZQQJRKFBP-WZRBSPASSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Securinine is a GABAA receptor antagonist which has been found to increase p73 protein expression
Securinine is an alkaloid widely in traditional folk medicine. Long known as a GABAA antagonist, securinine was recently found to up-regulate p53 protein and to modulate the related family member p73 protein in a p53-dependent fashion, inducing p73 in the HCT116 p53(-) cells and down-regulating it in the p53(+) cells. Induction of proapoptotic protein p73 only in p53 cells could be used to target cancer cells preferentially. Securinine has been found to induce p53-independent, p73-dependent apoptosis in RKO colon cancer cells.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service