Skip to Content
Merck
All Photos(1)

Key Documents

PHL83906

Sauchinone

phyproof® Reference Substance

Synonym(s):

(5aα,7α,8β,8aβ,14aS*,14bβ)-5a,6,7,8,8a,14b-Hexahydro-7,8-dimethyl-5H-benzo[kl]bis[1,3]dioxolo[4,5-b:4′,5′-g]xanthen-5-one, (5aR,7R,8S,8aR,14aS,14bR)-5a,6,7,8,8a,14b-Hexahydro-7,8-dimethyl-5H-Benzo[kl]bis[1,3]dioxolo[4,5-b:4′,5′-g]xanthen-5-one

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€385.00

€385.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€385.00

About This Item

Empirical Formula (Hill Notation):
C20H20O6
CAS Number:
Molecular Weight:
356.37
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€385.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

format

neat

storage temp.

2-8°C

SMILES string

O=C(C=C1OCO2)[C@]3([H])C[C@@H](C)[C@H](C)[C@@]4([H])C5=CC6=C(OCO6)C=C5O[C@]12[C@]43[H]

InChI

1S/C20H20O6/c1-9-3-11-13(21)5-17-20(25-8-24-17)19(11)18(10(9)2)12-4-15-16(23-7-22-15)6-14(12)26-20/h4-6,9-11,18-19H,3,7-8H2,1-2H3/t9-,10+,11+,18+,19+,20+/m1/s1

InChI key

GMTJIWUFFXGFHH-WPAOEJHSSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Sauchinone is an anti-inflammatory and antioxidant lignan isolated from Saururus chinensis a medical plant used for the treatment of fever, jaundice, oedema and inflammatory disease in oriental folk medicine. Sauchinone inhibits LPS-inducible iNOS (NOS II), TNF-α and COX-2 expression in macrophages through suppression of IKBα phosphorylation. It appears that sauchinone protects against myocardial ischemia/reperfusion injury by inhibition of p28 phosphorylation.
Sauchinone is an anti-inflammatory and antioxidant.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service