Skip to Content
Merck
All Photos(2)

Key Documents

371721

Sigma-Aldrich

GPR40 Agonist II

The GPR40 Agonist II controls the biological activity of GPR40. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

GPR40 Agonist II, 3-(4-((2,6-Dichloropyridin-4-yl)ethynyl)phenyl)propanoic acid, FFA1 Agonist II, Free Fatty Acid Receptor 1 Agonist II, FFA1 Agonist II, Free Fatty Acid Receptor 1 Agonist II, 3-(4-((2,6-Dichloropyridin-4-yl)ethynyl)phenyl)propanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€182.00

€182.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€182.00

About This Item

Empirical Formula (Hill Notation):
C16H11Cl2NO2
CAS Number:
Molecular Weight:
320.17
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

€182.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL, clear, colorless

shipped in

wet ice

storage temp.

2-8°C

SMILES string

ClC1=NC(Cl)=CC(C#CC2=CC=C(CCC(O)=O)C=C2)=C1

InChI

1S/C16H11Cl2NO2/c17-14-9-13(10-15(18)19-14)6-5-11-1-3-12(4-2-11)7-8-16(20)21/h1-4,9-10H,7-8H2,(H,20,21)

InChI key

GCWBKYDSSJIRNJ-UHFFFAOYSA-N

General description

A cell-permeable alkynedihydrocinnamic acid that acts as a potent GPR40/FFA1 agonist (EC50 = 40.7 nM in inducing Ca2+ mobilization in hFFA1-expressing 1321N1 cells) with >100-fold selectivity over GPR43/FFA2, GPR41/FFA3, and a panel of 56 other receptors, ion channels, and transporters. Reported to enhance 12 mM glucose-induced insulin secretion from rat INS-1E β-cells (by 34% at 10 µM) and exhibit good aqueous solubility (199 µM in PBS at pH 7.4), as well as chemical (0.1% degradation after 12 days in PBS at 37 °C) and metabolic (3% metabolization by human liver S9 stability test) stability.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C>

Other Notes

Christiansen, E., et al. 2011, J. Med. Chem.54, 6691.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service