Skip to Content
Merck
All Photos(1)

Key Documents

PHL80063

D,L-Camphene

phyproof® Reference Substance

Synonym(s):

Camphene

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€242.00

€242.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€242.00

About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€242.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (GC)

form

crystals

manufacturer/tradename

PhytoLab

bp

159-160 °C (lit.)

mp

48-52 °C (lit.)

density

0.85 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H][C@]12CC[C@]([H])(C1)C(C)(C)C2=C

InChI

1S/C10H16/c1-7-8-4-5-9(6-8)10(7,2)3/h8-9H,1,4-6H2,2-3H3/t8-,9+/m0/s1

InChI key

CRPUJAZIXJMDBK-DTWKUNHWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Other Notes

This compound is commonly found in plants of the genus: angelica cinnamomum curcuma eucalyptus mentha salvia thymus valeriana zingiber

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

78.8 °F - DIN 51755 Part 1

Flash Point(C)

26 °C - DIN 51755 Part 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service