Skip to Content
Merck
All Photos(1)

Key Documents

01410590

Camphene

primary reference standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€218.00

€218.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€218.00

About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€218.00


Check Cart for Availability

Request a Bulk Order

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

bp

159-160 °C (lit.)

mp

48-52 °C (lit.)

density

0.85 g/mL at 25 °C (lit.)

application(s)

food and beverages

SMILES string

[H][C@]12CC[C@]([H])(C1)C(C)(C)C2=C

InChI

1S/C10H16/c1-7-8-4-5-9(6-8)10(7,2)3/h8-9H,1,4-6H2,2-3H3/t8-,9+/m0/s1

InChI key

CRPUJAZIXJMDBK-DTWKUNHWSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

78.8 °F - DIN 51755 Part 1

Flash Point(C)

26 °C - DIN 51755 Part 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bénédicta G H Kpadonou Kpoviessi et al.
Journal of ethnopharmacology, 155(3), 1417-1423 (2014-07-25)
Different parts of Ocimum gratissimum Linn are largely used in folk medicine for the treatment of many diseases, some of which related to parasitical infections as fevers and headaches. In order to validate their use and to clarify the plant
Seon-Mi Seo et al.
Pesticide biochemistry and physiology, 113, 55-61 (2014-07-24)
This study investigated the fumigant toxicity of 4 Asteraceae plant essential oils and their constituents against the Japanese termite Reticulitermes speratus Kolbe. Fumigant toxicity varied with plant essential oils or constituents, exposure time, and concentration. Among the tested essential oils
M Melusova et al.
Neoplasma, 61(6), 690-699 (2014-10-25)
For several thousand years natural products were successfully used to treat a variety of diseases and to maintain health in humans, but until now it is not fully known what causes these medicinal effects. In our study we assessed the cytotoxic
Karine Rech Begnini et al.
Journal of medicinal food, 17(10), 1129-1133 (2014-09-18)
Cancer is a leading cause of death and is responsible for one in eight deaths worldwide. The use of herbs as complementary medicine for cancer, especially advanced cancer, has recently increased. The aim of this study was to evaluate in
Luana Quassinti et al.
Fitoterapia, 97, 133-141 (2014-06-14)
Smyrnium olusatrum (Apiaceae), well known as wild celery, is a biennal celery-scented plant used for many centuries as a vegetable, then abandoned after the introduction of celery. In the present work, the essential oil obtained from inflorescences and the amounts

Protocols

GC Analysis of Sweet Orange Essential Oil on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service