Skip to Content
Merck
All Photos(1)

Key Documents

MET12133A

Supelco

(Aminomethyl)phosphonic acid

analytical standard

Synonym(s):

AMPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2P(O)(OH)2
CAS Number:
Molecular Weight:
111.04
Beilstein:
1701219
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. MET-12133A-100MG

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

NCP(O)(O)=O

InChI

1S/CH6NO3P/c2-1-6(3,4)5/h1-2H2,(H2,3,4,5)

InChI key

MGRVRXRGTBOSHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Börjesson et al.
Journal of chromatography. A, 886(1-2), 207-216 (2000-08-19)
New methods were developed to determine glyphosate, N-(phosphonomethyl)glycine, and its major metabolite, (aminomethyl)phosphonic acid in groundwater and soil. The methods involve ligand-exchange, anion-exchange and derivatisation and final identification and quantification by GC-MS. The limits of quantification in this experiment were
G M Williams et al.
Regulatory toxicology and pharmacology : RTP, 31(2 Pt 1), 117-165 (2000-06-15)
Reviews on the safety of glyphosate and Roundup herbicide that have been conducted by several regulatory agencies and scientific institutions worldwide have concluded that there is no indication of any human health concern. Nevertheless, questions regarding their safety are periodically
M Tomita et al.
Journal of chromatography, 571(1-2), 324-330 (1991-11-15)
Capillary electrophoresis has been used to separate and quantitate glyphosate and its major metabolite, (aminomethyl)phosphonic acid (AMPA), in serum. The two compounds, after derivatization with p-toluenesulphonyl chloride, were clearly separated with 0.1 M boric acid-sodium hydroxide buffer (pH 9.6) containing
Chuanmin Ruan et al.
Analytica chimica acta, 605(1), 80-86 (2007-11-21)
Techniques for rapid screening of uranium in environmental samples are needed, and this study entails the development of surface-enhanced Raman scattering (SERS) for analyzing uranium in aqueous media with improved sensitivity and reproducibility. A new SERS substrate based on (aminomethyl)phosphonic
M E Oppenhuizen et al.
Journal - Association of Official Analytical Chemists, 74(2), 317-323 (1991-03-01)
A new method for determination of glyphosate and aminomethylphosphonic acid (AMPA) residues in environmental water was collaboratively studied by 6 laboratories. The method is simpler and shorter than previous methods. A filtered volume of water is evaporated to dryness and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service