Skip to Content
Merck
All Photos(1)

Key Documents

CRM67124

Supelco

Methiocarb sulfone

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Mercaptodimethursulfon, Methiocarb sulfone

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€262.00

€262.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€262.00

About This Item

Empirical Formula (Hill Notation):
C11H15NO4S
CAS Number:
Molecular Weight:
257.31
Beilstein:
2291460
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€262.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

SMILES string

CNC(=O)Oc1cc(C)c(c(C)c1)S(C)(=O)=O

InChI

1S/C11H15NO4S/c1-7-5-9(16-11(13)12-3)6-8(2)10(7)17(4,14)15/h5-6H,1-4H3,(H,12,13)

InChI key

RJBJMKAMQIOAML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Methiocarb sulfone is a carbamate pesticide and one of the four metabolites of insecticide, molluscicide, and bird-repellent methiocarb.
As per the commission implementing regulation (EU) 2019/1606 of 27th September 2019, methiocarb (expressed as the sum of methiocarb, methiocarb sulfone, and methiocarb sulfoxide) is not approved for use in the European Union. Maximum residue levels (MRLs) have been set according to Reg (EC) No 149/2008 for methiocarb (expressed as the sum of methiocarb, methiocarb sulfone, and methiocarb sulfoxide) for various products of plant and animal origin from 0.01 to 0.1 mg/kg. Methiocarb has to be monitored in the Multiannual Control Program for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
The methiocarb sulfone CRM may also find following uses:
  • Method development for simultaneous determination of methiocarb, methiocarb sulfoxide, and methiocarb sulfone in five food products of animal origin (milk, chicken, pork, beef, and eggs) by liquid chromatography-tandem mass spectrometry (LC-MS/MS)
  • Determination of 304 pesticide residues in citrus fruit (grapefruit, lemon, orange and mandarin) and vegetable (cucumber, eggplant, tomato and zucchini) samples by liquid chromatography-tandem mass spectroscopy (LC-MS/MS)
  • Analysis of environmental water samples for the presence of 23 carbamate pesticide residues by solid-phase extraction (SPE) combined with ultra-high-performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)
  • Develop a UHPLC-MS/MS method to determine 28 carbamate pesticide residues in fresh herb samples of basil, spearmint, wild mint, parsley, Chinese parsley, thyme, chives, and salvia after their QuEChERS-based extraction
  • Validation of a method based on QuEChERS extraction and LC-MS/MS and GC-MS/MS quantification of 218 pesticide residues in clay loam soil
  • Multi-residue analysis of pesticides and mycotoxins in red wine using direct injection for sample preparation and UPLC-MS/MS for analysis
  • Method development for detection and quantification of 260 pesticides and mycotoxins in green tea and royal jelly nutraceutical products by “dilute and shoot” extraction procedure and quantification by LC-Orbitrap-MS

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Acosta-Dacal et al.
Data in brief, 33, 106393-106393 (2020-11-05)
The dataset presented in this article supports "Optimization and validation of a method for the simultaneous environmental monitoring of 218 pesticide residues in clay loam soil" [1]. A method based on QuEChERS (Quick, Easy, Cheap, Effective, Rugged & Safe) for
Gerardo Martínez-Domínguez et al.
Food chemistry, 197(Pt A), 907-915 (2015-12-01)
A multi-class methodology was developed to determine pesticides and mycotoxins in food supplements. The extraction was performed using acetonitrile acidified with formic acid (1%, v/v). Different clean-up sorbents were tested, and the best results were obtained using C18 and zirconium
Jonatan V Dias et al.
Food chemistry, 293, 83-91 (2019-06-04)
A simple and rapid method for simultaneous determination of pesticides and mycotoxins in red wine is presented. Sample preparation approach, called direct injection, consists of a sequence of only three steps: centrifugation, dilution and filtration. The analysis of extracts were
Characterization of Carbamate Pesticides in Natural Water from Cameroon
Nantia EA, et al.
Analytical Letters, 50(9) (2017)
Edouard Akono Nantia et al.
Food chemistry, 216, 334-341 (2016-09-07)
A new reliable, fast and highly sensitive method based on ultra-high performance liquid chromatography tandem mass spectrometry has been developed and validated for the determination of 28 carbamates in aromatic herbs. A modified QuEChERS-based method was optimized for the extraction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service