Skip to Content
Merck
All Photos(1)

Documents

34177

Supelco

Methiocarb sulfoxide

PESTANAL®, analytical standard

Synonym(s):

4-Methylsulfinyl-3,5-xylyl-N-methylcarbamate, Mercaptodimethur sulfoxide, Mesurol sulfoxide, Methiocarb breakdown product

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO3S
CAS Number:
Molecular Weight:
241.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CNC(=O)Oc1cc(C)c(c(C)c1)S(C)=O

InChI

1S/C11H15NO3S/c1-7-5-9(15-11(13)12-3)6-8(2)10(7)16(4)14/h5-6H,1-4H3,(H,12,13)

InChI key

FNCMBMZOZQAWJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methiocarb sulfoxide, is a carbamate insecticide, which is used to control rice insects. It is a metabolite of methiocarb present in honey.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas chromatographic analysis of methiocarb and its metabolites in soil and rice
Keum.S.Y, et al.
Chromatographia, 52, 237-241 (2000)
Development and validation of a multi-residue method for pesticide determination in honey using on-column liquid?liquid extraction and liquid chromatography?tandem mass spectrometry
Pirard C, et al.
Journal of Chromatography A, 1152, 116-123 (2007)
Md Musfiqur Rahman et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1060, 387-394 (2017-07-01)
A simultaneous analytical method was developed for the determination of methiocarb and its metabolites, methiocarb sulfoxide and methiocarb sulfone, in five livestock products (chicken, pork, beef, table egg, and milk) using liquid chromatography-tandem mass spectrometry. Due to the rapid degradation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service