Skip to Content
Merck
All Photos(1)

Key Documents

94057

Supelco

Bicyclohexyl

analytical standard

Synonym(s):

Dicyclohexyl

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€40.50

€40.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€40.50

About This Item

Linear Formula:
C6H11C6H11
CAS Number:
Molecular Weight:
166.30
Beilstein:
1848266
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€40.50


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

form

liquid

autoignition temp.

473 °F

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.478 (lit.)

bp

227 °C (lit.)

mp

3-4 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

SMILES string

C1CCC(CC1)C2CCCCC2

InChI

1S/C12H22/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h11-12H,1-10H2

InChI key

WVIIMZNLDWSIRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Asp. Tox. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service