Skip to Content
Merck
All Photos(1)

Key Documents

89407

Supelco

BDE No 209 solution

~50 μg/mL (Isooctane:Toluene 50:50), analytical standard

Synonym(s):

Decabromodiphenyl ether solution, Decabromodiphenyl oxide

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€208.00

€208.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€208.00

About This Item

Empirical Formula (Hill Notation):
C12Br10O
CAS Number:
Molecular Weight:
959.17
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€208.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

form

solution

shelf life

limited shelf life, expiry date on the label

concentration

~50 μg/mL (Isooctane:Toluene 50:50)

storage temp.

2-8°C

SMILES string

Brc1c(Br)c(Br)c(Oc2c(Br)c(Br)c(Br)c(Br)c2Br)c(Br)c1Br

InChI

1S/C12Br10O/c13-1-3(15)7(19)11(8(20)4(1)16)23-12-9(21)5(17)2(14)6(18)10(12)22

InChI key

WHHGLZMJPXIBIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

The customer needs to make sure that the standard is homogeneous, this can be achieved by shaking, vortexing , or sonicating the ampoule before opening the ampoule. Care should be taken when opening the ampoule to avoid any injuries from the broken glass.
The standard should be transferred to a clean and appropriate vial or flask using clean pipettes or micro pipettes. The vial should be immediately capped to avoid any loss or evaporation of the solvent.
After opening the ampoule, the standard should not be stored or kept in the ampoule. To preserve the integrity of the product, the standard should be transferred to an appropriate vial that must be capped and stored according to the recommendation on the Certificate of Analysis.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service