Skip to Content
Merck
All Photos(1)

Key Documents

525189

Sigma-Aldrich

Dioctyl terephthalate

≥96%

Synonym(s):

Bis(2-ethylhexyl) terephthalate

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€68.20
20 L
€521.00

€68.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€68.20
20 L
€521.00

About This Item

Linear Formula:
C6H4-1,4-[CO2CH2CH(C2H5)(CH2)3CH3]2
CAS Number:
Molecular Weight:
390.56
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€68.20


Check Cart for Availability

Request a Bulk Order

vapor pressure

1 mmHg ( 217 °C)

Quality Level

Assay

≥96%

refractive index

n20/D 1.49 (lit.)

bp

400 °C (lit.)

mp

−48 °C

density

0.986 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)COC(=O)c1ccc(cc1)C(=O)OCC(CC)CCCC

InChI

1S/C24H38O4/c1-5-9-11-19(7-3)17-27-23(25)21-13-15-22(16-14-21)24(26)28-18-20(8-4)12-10-6-2/h13-16,19-20H,5-12,17-18H2,1-4H3

InChI key

RWPICVVBGZBXNA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dioctyl terephthalate (DOTP) is a plasticizer that can be prepared by alcoholysis of polyethylene terephthalate (PET) with isooctyl alcohol.[1]

Application

DOTP can form a composite blend with polyaniline (PANI)-ethyene dimethacrylate (EDMA), which can be used in the fabrication of conductive films.[2][3] It may also be used as a plasticizer with polyvinyl chloride (PVC) to form solutions for artificial intravessel plaque.[4]

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

413.6 °F - closed cup

Flash Point(C)

212 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alcoholysis of PET to produce dioctyl terephthalate by isooctyl alcohol with ionic liquid as cosolvent
Chen J, et al.
Polymer Degradation and Stability, 107, 178-183 (2014)
Effect of ferric chloride on the electrical conductivity and characterization of polyethylene oxide/polyvinyl chloride/polyaniline conductive films
AIP Conference Proceedings, 1985(1), 020002-020002 (2018)
Enhancement of Electrical Conductivity and Tensile Properties of Conductive Poly (vinyl Chloride)/Poly (ethylene Oxide)/Polyaniline Conductive Composite Films: Effect of Polyaniline Loading and Ethylene Dimethacrylate.
Yazid MIM, et al.
Journal of Physical Science, 29(2), 178-183 (2018)
D C Topping et al.
Toxicology and industrial health, 3(2), 63-78 (1987-06-01)
Groups of five male and five female rats were fed diets containing from 0% to 2.5% di(2-ethylhexyl)terephthalate (DEHT) or 1.2% di(2-ethylhexyl)phthalate (DEHP) for 21 days. Feed consumption and body weight gains were collected and, at study termination, animals were examined
E D Barber
Environmental and molecular mutagenesis, 23(3), 228-233 (1994-01-01)
Di(2-ethylhexyl) terephthalate (DEHT) is a commercially produced chemical (Kodaflex DOTP) that is used as a general purpose, low-volatility plasticizer for polyvinyl chloride and other polymeric materials. Less than 30 million kilograms of DEHT are produced annually. DEHT is isomeric with

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service