Skip to Content
Merck
All Photos(1)

Key Documents

89125

Supelco

Suberic acid-2,3,4,5,6,7-13C6

analytical standard

Synonym(s):

Octanedioic acid-2,3,4,5,6,7-13C6, Suberic acid-13C6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C2H14O4
Molecular Weight:
180.15
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

form

powder

shelf life

limited shelf life, expiry date on the label

extent of labeling

≥99%

storage temp.

2-8°C

SMILES string

OC([13CH2][13CH2][13CH2][13CH2][13CH2][13CH2]C(O)=O)=O

InChI

1S/C8H14O4/c9-7(10)5-3-1-2-4-6-8(11)12/h1-6H2,(H,9,10)(H,11,12)/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

TYFQFVWCELRYAO-IDEBNGHGSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service