Skip to Content
Merck
All Photos(1)

Key Documents

457086

Sigma-Aldrich

Phthalic anhydride-d4

98 atom % D

Synonym(s):

1,3-Isobenzofurandione-4,5,6,7-d4

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€605.00

€605.00


Check Cart for Availability

Request more information

Select a Size

Change View
500 MG
€605.00

About This Item

Empirical Formula (Hill Notation):
C8D4O3
CAS Number:
Molecular Weight:
152.14
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

€605.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % D

form

solid

bp

284 °C (lit.)

mp

131-134 °C (lit.)

mass shift

M+4

SMILES string

[2H]c1c([2H])c([2H])c2C(=O)OC(=O)c2c1[2H]

InChI

1S/C8H4O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h1-4H/i1D,2D,3D,4D

InChI key

LGRFSURHDFAFJT-RHQRLBAQSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service