Skip to Content
Merck
All Photos(1)

Key Documents

821464

TSKgel® BioAssist® Desalting (13 µm) HPLC Columns

L x I.D. 15 cm × 10 mm, polyacrylamide phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
Pricing and availability is not currently available.

Product Name

TSKgel® BioAssist® Desalting HPLC Column, phase polyacrylamide, L × I.D. 15 cm × 10 mm, 13 μm particle size, PEEK

material

PEEK

product line

TSKgel®

manufacturer/tradename

Tosoh 21464

parameter

≤30% polar organic solvent
13 bar max. pressure
4 mL/min flow rate
4-30 °C temperature

technique(s)

HPLC: suitable

L × I.D.

15 cm × 10 mm

matrix

urea cross-linked polyacrylamide

matrix active group

polyacrylamide phase

particle size

13 μm

operating pH

3.0-12.0(<= 1 wk)
4.0-11.0(> 1 wk)

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel BioAssist DS columns are designed for the desalting and buffer exchange of proteins and polynucleotides at semi-preparative scale. Packed with 15μm polyacrylamide beads in PEEK hardware, TSKgel BioAssist DS columns show excellent desalting performance.
The novel, hydrophilic, highly cross-linked polyacrylamide beads exhibit superior mechanical strength compared with conventional hydrophilic polyacrylamide beads and cross-linked dextran beads. This increase in strength is what allows the use of the small spherical 15μm beads. TSKgel BioAssist DS columns can be operated in standard HPLC systems to quickly and efficiently reduce salt and/or buffer concentrations of collected protein or nucleic acid fractions.

Physical form

Shipped in water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

BioAssist is a registered trademark of Tosoh Corporation
TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service