Skip to Content
Merck
All Photos(1)

Documents

5037804

Supelco

Amplifex Diene Reagent Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
NA.21

technique(s)

LC/MS: suitable

Quality Level

shipped in

dry ice

storage temp.

−70°C

General description

Kit contains a protocol, 1 tubes of Amplifex Diene reagent and 1 tube of diluent. Each tube contains enough reagent to complete 50 reactions.

Analysis Note

For further information on the Amplifex Diene Reagent kit, please visit this link.

To view the Protocol for the Amplifex Diene Reagent Kit, please visit this Protocol link.

Additional Application Notes may be viewed at the following links:
* Sample Preparation Procedure and Amplifex Diene Reagent Chemistry for DHVD3/DHVD2 Quantitation
* Analysis of 1,25 DHVD3 and 1,25 DHVD2 Using Amplifex Diene Reagent

Legal Information

Amplifex is a trademark of AB Sciex Pte. Ltd.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F

Flash Point(C)

2.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service