Skip to Content
Merck
All Photos(1)

Key Documents

817396

TOYOPEARL® AF-Formyl-650M Bulk Media

phase aldehyde, bottle of 1 L, 65 μm particle size

Synonym(s):

TOYOPEARL® AF-Formyl-650M Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
particle size:
65 μm
matrix active group:
aldehyde phase
pore size:
1000 Å
matrix:
hydroxylated methacrylic polymer particle platform
Pricing and availability is not currently available.

description

phase filter (Formyl)

product line

TOYOPEARL®

form

aqueous slurry in 1% glutaraldehyde

packaging

bottle of 1 L

manufacturer/tradename

Tosoh 17396

extent of labeling

40-70 μeq/mL ligand density

parameter

pressure (max. 3 bar)

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer particle platform

matrix active group

aldehyde phase

particle size

65 μm

pore size

1000 Å

pH

6-9

separation technique

affinity

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl AF-Formyl-650 resin is a reactive resin which binds exclusively to primary amines to form a stable secondary amine linkage.

Physical form

Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service