Skip to Content
Merck
All Photos(1)

Key Documents

779164

Sigma-Aldrich

2-Azidotoluene solution

~0.5 M in tert-butyl methyl ether, ≥95.0% (HPLC)

Synonym(s):

1-Azido-2-methylbenzene solution, 2-Methylphenyl azide solution, o-Tolyl azide solution

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€208.00

€208.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
€208.00

About This Item

Empirical Formula (Hill Notation):
C7H7N3
CAS Number:
Molecular Weight:
133.15
Beilstein:
1447887
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

€208.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95.0% (HPLC)

form

liquid

concentration

~0.5 M in tert-butyl methyl ether

impurities

≤2.0% water

storage temp.

−20°C

SMILES string

Cc1ccccc1N=[N+]=[N-]

InChI

1S/C7H7N3/c1-6-4-2-3-5-7(6)9-10-8/h2-5H,1H3

InChI key

ZKGVLNVASIPVAU-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service