Skip to Content
Merck
All Photos(1)

Key Documents

77068

Supelco

Thiofanox

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€92.40

€92.40


Estimated to ship on25 April 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€92.40

About This Item

Empirical Formula (Hill Notation):
C9H18N2O2S
CAS Number:
Molecular Weight:
218.32
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€92.40


Estimated to ship on25 April 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

−20°C

SMILES string

CNC(ON=C(C(C)(C)C)CSC)=O

InChI

1S/C9H18N2O2S/c1-9(2,3)7(6-14-5)11-8(12)13-10-4/h10H,6H2,1-5H3/b11-7+

InChI key

AGBTZYDUBVIKEZ-YRNVUSSQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Thiofanox is a systemic insecticide and acaricide that belongs to the group of oxime carbamates. It is an acetylcholinesterase (AChE) inhibitor causing hyperexcitation and impeding the functioning of the nervous system. Thiofanox is used for protection against black bean aphids, beet leaf miners, flea beetles, and pygmy mangold beetles in beets; flea beetles in rape; aphids, capsids, flea beetles, Colorado beetles, and leafhoppers in potatoes; aphids, midges, flea beetles, soil-inhabiting larvae, and spider mites in cotton and aphids; and spider mites in floriculture.
According to the Commission Implementing Regulation (EU) No 2076/2002 of 20th November 2002, thiofanox is not approved for use in the European Union (EU) as a plant protection agent, following the Regulation (EC) No. 1107/2009. But a default maximum residue limit of 0.01 mg/kg has been set for the presence of thiofanox in products of plant and animal origin, according to Art 18(1)(b) Reg 396/2005.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Thiofanox CRM may also be used as given below:

  • 126 pesticide residues in chicken samples using modified QuEChERS for sample pre-treatment and ultra-high-performance liquid chromatography combined with quadrupole time-of-flight mass spectrometry (UHPLC-QToF-MS) for quantification
  • Multi-residue analysis of 423 pesticides, consisting of their isomers and metabolites in 124 green tea (C. sinensis) nutraceutical products by two-dimensional gas chromatography coupled with time-of-flight mass spectrometry
  • QuEChERS based extraction and ultra-high performance liquid chromatography-triple quadrupole tandem mass spectrometric (UHPLC-QqQ-MS/MS) analysis of LC-amenable pesticides in green tea (C. sinensis) based nutraceutical products
  • Multi-residue analysis of 80 pesticide residues in 30 aquaculture water samples by single step solid phase extraction (SPE)-cleanup and ultra-high performance liquid chromatography-Q/Orbitrap mass spectrometry

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service