Skip to Content
Merck
All Photos(1)

Key Documents

57042

Supelco

Visprep SPE Manifold Test Tubes, 10 x 75 mm

pk of 12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
Pricing and availability is not currently available.

packaging

pk of 12

Quality Level

technique(s)

solid phase extraction (SPE): suitable

size

10 mm × 75 mm

application(s)

food and beverages

General description

Visprep SPE manifold test tubes, 10 x 75 mm are technically used for a 12-port or 24-port Visiprep SPE vacuum manifold.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service