Documents
57044
Supelco
Visiprep SPE Vacuum Manifold
DL (Disposable Liner), 12-port model
About This Item
UNSPSC Code:
12000000
material
polypropylene tube
technique(s)
solid phase extraction (SPE): suitable
application(s)
environmental
food and beverages
General description
The Visiprep DL (Disposable Liner) Vacuum Manifold eliminates the possibility of cross-contamination when processing a new sample on the same port. The liner consists of a PP female luer hub that attaches to the SPE, and thin-walled PTFE tubing that is threaded through the SPE port. This ensures that all SPE port and valve surfaces coming in contact with the sample can be replaced following each extraction.
SPE is a form of chromatography, and, as with other chromatographic techniques, control of flow rate is critical for maintaining reproducible extractions. Unlike other vacuum manifolds, the Visiprep system contains a patented valve system that allows for precise flow control through each SPE tube via rotating, independent, screw-type valves situated in each port within the manifold cover. Visiprep vacuum manifolds allow you to process up to 12 (12-port version) or 24 (24-port version) SPE samples simultaneously. Spacing between ports are slightly smaller on the 24-port versions than the 12-port versions so consideration of tube sizes is recommended.
Features and Benefits
  • Patented screw-type valves within each SPE port for precise flow control
  • Glass basin will not dissolve, fog, or discolor when exposed to solvents
  • Legs on cover allow user to easily rest cover on work surface when removed from the manifold
  • Screw-type solvent-resistant vacuum bleed gauge and valve offer better sealing and vacuum control. Valve takes 1/4″ vacuum tubing.
  • PP collection vessel rack accommodates autosampler vials; small scintillation vials (22.75 mm O.D. recommended); 10 and 16 mm test tubes; and 1, 2, 5, and 10 mL volumetric flasks. An optional plate for 20 mL scintillation vials is available for 12-port models.
Supplied with 100 disposable liners (Part No. 57059).
Legal Information
Visiprep is a trademark of Sigma-Aldrich Co. LLC
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Andrea Molina et al.
Toxins, 11(6) (2019-06-05)
Mycotoxins are secondary metabolites, produced by fungi of genera Aspergillus, Penicillium and Fusarium (among others), which produce adverse health effects on humans and animals (carcinogenic, teratogenic and immunosuppressive). In addition, mycotoxins negatively affect the productive parameters of livestock (e.g., weight
Articles
Use QuEChERS methods to analyze samples of milk for veterinary drugs.
Dual-layer cartridge achieves efficient SPE clean-up for challenging pesticide residue testing in turmeric and spices.
2-layer SPE cartridge efficiently reduces background for ginger contaminant analysis.
Analysis of 20 pharmaceutical compounds in calf serum using Supel™ Swift HLB SPE cartridge for cleanup and LC-MS determination and another commercially available HLB cartridge for comparison.
Protocols
Paprika spice's PAH contamination risk due to environmental exposure highlights food safety concerns.
Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.
Related Content
Solid phase extraction purifies samples for chromatographic analysis, improving data quality and protecting analytical columns.