Skip to Content
Merck
All Photos(1)

Key Documents

54808

Sigma-Aldrich

Mono S® 5/50 GL

10 μm particle size, L × I.D. 5 cm × 5 mm

Synonym(s):

Amersham Biosciences Ion Exchange Columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817

L × I.D.

5 cm × 5 mm

matrix

MonoBeads

matrix active group

sulfonic acid

particle size

10 μm

compatibility

mode of use ion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Mono columns are highly efficient, pH-stable columns designed for high performance ion exchange separations of proteins, peptides, and polynucleotides. The unique properties of these columns are based on MonoBeads support, a beaded hydrophilic material with the narrowest particle size distribution of any chromatographic support. This monodispersity permits high flow rates at relatively low backpressures.

Application

Mono columns are used in applications including peptide mapping and monoclonal antibody purification. Mono S columns have been used in cancer research studies.

Legal Information

Mono S is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiu-Juan Ye et al.
Journal of agricultural and food chemistry, 59(18), 10232-10238 (2011-08-13)
A 30 kDa antifungal protein was purified from red cabbage ( Brassica oleracea ) seeds. It exhibited a molecular mass and N-terminal amino acid sequence disinct from those of previously isolated Brassica antifungal proteins. The protocol used entailed ion exchange
Evandro Fei Fang et al.
Archives of toxicology, 86(2), 293-304 (2011-10-05)
A 20-kDa Kunitz-type trypsin-chymotrypsin inhibitor, Bauhinia purpurea trypsin inhibitor (BPLTI), has been isolated from the seeds of B. purpurea L. by using liquid chromatography procedures that involved ion exchange chromatography on Sp-Sepharose and Mono S and gel filtration on Superdex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service