Merck
HPLC Analysis of White-Faced Hornet Venom on Mono S® HR5/5
HPLC Analysis of White-Faced Hornet Venom on Mono S® HR5/5 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Mono S® 5/50 GL
10 μm particle size, L × I.D. 5 cm × 5 mm
CONDITIONS
column
Mono S HR5/5, 10 μm particles (54808)
mobile phase
[A] 50 mM BICINE, pH 8.4; [B] A + 0.35 M sodium chloride
flow rate
1 mL/min
detector
UV, 280 nm
injection
7 mg venom in mobile phase A
Description
Legal Information
Mono S is a registered trademark of Cytiva