Skip to Content
Merck
All Photos(1)

Documents

51104AST

Supelco

Astec® Cellulose DMP Chiral (5 μm) HPLC Column

L × I.D. 2 cm × 2.1 mm, , HPLC Guard Cartridge, pkg of 2 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

product name

Astec® Cellulose DMP Chiral HPLC Guard, Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm, pkg of 2 ea

material

stainless steel column

Agency

suitable for USP L40

product line

Astec®

packaging

pkg of 2 ea

technique(s)

HPLC: suitable

L × I.D.

2 cm × 2.1 mm

matrix

fully porous particle
silica gel particle platform

matrix active group

cellulose, DMPC- phase

particle size

5 μm

pore size

100 Å pore size

operating pH

3.5-6.8

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

General description

Guard cartridges for Astec® Cellulose DMP, 2.1 mm I.D. columns. Requires holder 59660-U.
Guard cartridges to protect Astec® Cellulose DMP columns. Choose the I.D. to match the I.D. of your analytical column. The 2.1 mm and 4 mm I.D. cartridges are sold in kits that contain the holder and fittings, and as packs of 2 cartridges. The 10 mm and 21.2 mm I.D. guards are sold as individual cartridges that require holders, sold separately.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Separate a wide range of chiral compounds via normal phase, polar organic, and SFC conditions; useful in pharma, chemical, & environmental industry applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service