Skip to Content
Merck
All Photos(1)

Key Documents

1.51486

Supelco

MANU-CARTNT HPLC cartridge holder

for LiChroCART 2,3,4,4.6 mm i.d. HPLC cartridges

Synonym(s):

TM=["MANU-CART"] holder for TM=["LICHROCART"] HPLC-cartridges

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€562.00

€562.00


Estimated to ship on24 May 2025



Select a Size

Change View
1 UNIT
€562.00

About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

€562.00


Estimated to ship on24 May 2025


Product Name

manu-CART NT cartridge holder for LiChroCART 2,3,4,4.6 mm i.d. HPLC cartridges,

technique(s)

HPLC: suitable

storage temp.

no temp limit

General description

manu-CART NT cartridge holder for 2, 3, 4 and 4.6 mm i.d. LiChroCART cartridges.

Analysis Note

visual control: passes test

Legal Information

LICHROCART is a trademark of Merck KGaA, Darmstadt, Germany
MANU-CART is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Can you provide guidance on selecting the appropriate fittings for this column?

    1 answer
    1. LiChroCART HPLC columns are designed in a "cartridge" style and do not include end fittings. The appropriate end fittings for LiChroCART HPLC columns are the manu-CART® NT cartridge holder for LiChroCART® 2, 3, 4, 4.6 mm i.d. HPLC cartridges, product 1.51486.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service