Skip to Content
Merck
All Photos(1)

Key Documents

4M4829-U

Supelco

1,4-Dioxane-d8

certified reference material, 2000 μg/mL in methanol, ampule of 5 × 1 mL

Synonym(s):

p-Dioxane-d8, Octadeuterodioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4D8O2
CAS Number:
Molecular Weight:
96.15
Beilstein:
1307286
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 5 × 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.4198 (lit.)

bp

99 °C (lit.)

density

1.129 g/mL at 25 °C (lit.)

application(s)

environmental

format

single component solution

mass shift

M+8

storage temp.

2-8°C

SMILES string

[2H]C1([2H])OC([2H])([2H])C([2H])([2H])OC1([2H])[2H]

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2/i1D2,2D2,3D2,4D2

InChI key

RYHBNJHYFVUHQT-SVYQBANQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leah E Jackson et al.
Journal of separation science, 44(4), 860-869 (2020-12-17)
In this study, we developed an analytical method for the determination of 1,4-dioxane in aqueous solutions using freeze-assisted liquid-liquid extraction, also known as frozen microextraction, and gas chromatography with triple quadrupole mass spectrometry with select reaction monitoring. The method is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service