Skip to Content
Merck
All Photos(1)

Key Documents

48701

Supelco

Aroclor 1016 solution

certified reference material, 200 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Agency

EPA 505,508,508A,8080,8082,508.1
MISA (Canada) Group 27

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

InChI

1S/C12H7Cl3/c13-10-3-1-2-8(4-10)9-5-11(14)7-12(15)6-9/h1-7H

InChI key

RIBGNAJQTOXRDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Flam. Liq. 2 - STOT RE 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sofia Abassi et al.
Cell stress & chaperones, 25(6), 1117-1123 (2020-08-15)
Heat shock proteins (HSPs) and HSP70-HSP90 organizing proteins (HOPs) are related, and they function together to maintain cellular homeostasis and respond to stress. In the present study, we reported the first molecular characteristics of HSP70 (designated as CeHSP70) and HOP
Monika G Baldridge et al.
Reproductive toxicology (Elmsford, N.Y.), 17(5), 567-573 (2003-10-14)
Polychlorinated biphenyls (PCBs) are ubiquitous man-made toxicants capable of endocrine disruption. Studies in several species have shown that exposure to PCBs and their hydroxylated metabolites reduces fecundity and decreases circulating concentrations of thyroid hormones, causing serious reproductive and developmental defects.
R F Seegal et al.
Toxicology, 66(2), 145-163 (1991-02-01)
Adult male non-human primates, Macaca nemestrina, were orally-exposed to corn oil or corn oil containing either Aroclor 1016 or 1260 at doses of 0.8, 1.6 or 3.2 mg/(kg.day) for 20 weeks. Brain concentrations of biogenic amines and individual PCB congeners
R W Lawton et al.
Archives of environmental health, 40(1), 29-37 (1985-01-01)
The 95% prediction interval for single measurements of serum "Aroclor" reported by a reputable commercial analyst was found to be approximately +/- 42%. The geometric mean serum PCB levels in a population of capacitor workers who had formerly had direct
Marika Viisimaa et al.
Environmental technology, 33(16-18), 2041-2048 (2012-12-18)
The potential and feasibility of treating soil contaminated with electrical insulating oil, Aroclor 1016, containing polychlorinated biphenyls (PCBs) with stabilized hydrogen peroxide were evaluated using columns packed with soils of two different matrixes. The column experiments showed that PCBs degraded

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service