Skip to Content
Merck
All Photos(1)

Key Documents

48530-U

Supelco

1,3-Dichloropropene

cis + trans, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH=CHCl
CAS Number:
Molecular Weight:
110.97
Beilstein:
1719556
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 2000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.472 (lit.)

bp

97-112 °C (lit.)

density

1.198 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

ClC\C=C/Cl

InChI

1S/C3H4Cl2/c4-2-1-3-5/h1-2H,3H2/b2-1+

InChI key

UOORRWUZONOOLO-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Dichloropropene is a chlorinated alkene with a chloroform-like smell. It finds application as a preplanting fumigant against nematodes harming the plant roots, wireworms, weeds, etc. It can also be used as an intermediate in the preparation of several pesticides.[1]

Application

1,3-Dichloropropene may be used as an analytical reference standard for the determination of the analyte in air, water and soil samples by various chromatography techniques.[2][3][4]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Static headspace and gas chromatographic analysis of fumigant residues in soil and water
Gan J, et al.
Journal of Agricultural and Food Chemistry, 46(3), 986-990 (1998)
Pesticide analysis by high-performance liquid chromatography using the direct injection method
Kiso Y, et al.
Journal of Chromatography A, 733(1-2), 259-265 (1996)
NTP 12th report on carcinogens.
Report on Carcinogens (12th Ed. ), 12(1-2), iii-iii (2011)
Development and validation of an air monitoring method for 1, 3-dichloropropene, trans-1, 2, 3-trichloropropene, cis-1, 2, 3-trichloropropene, 1, 1, 2, 3-tetrachloropropene, 2, 3, 3-trichloro-2-propen-1-ol, and 1, 1, 2, 2, 3-pentachloropropane
Leiber MA and Berk HC
Analytical Chemistry, 56(12), 2134-2137 (1984)
1,3-Dichloropropene (technical grade).
Report on carcinogens : carcinogen profiles, 12, 150-152 (2011-08-20)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service