Skip to Content
Merck
All Photos(1)

Key Documents

48196

Supelco

EPA Pesticide Standard Mix B

certified reference material, in hexane: toluene (99:1) (varied)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Agency

EPA

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type pesticide/PCB

packaging

ampule of 1 mL

concentration

in hexane: toluene (99:1) (varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Aldrin 5 μg/mL
    β-BHC
    δ-BHC
    α-Chlordane 5 μg/mL
    γ-Chlordane 5 μg/mL
    1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene 10 μg/mL
    β-Endosulfan 10 μg/mL
    Endosulfan sulfate 10 μg/mL
    Endrin aldehyde 10 μg/mL
    Endrin ketone 10 μg/mL
    Heptachlor exo-epoxide 5 μg/mL
    See All (11)

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service