Skip to Content
Merck
All Photos(1)

Key Documents

47956

Supelco

Permethrin (isomers) solution

cis/trans isomers, 1000 μg/mL in methanol, PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
5765325
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

packaging

ampule of 1 mL

concentration

1000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

room temp

SMILES string

CC1(C)C(\C=C(\Cl)Cl)C1C(=O)OCc2cccc(Oc3ccccc3)c2

InChI

1S/C21H20Cl2O3/c1-21(2)17(12-18(22)23)19(21)20(24)25-13-14-7-6-10-16(11-14)26-15-8-4-3-5-9-15/h3-12,17,19H,13H2,1-2H3

InChI key

RLLPVAHGXHCWKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Permethrin (isomers) solution may be used as a reference standard for the determination of permethrin in pharmaceutical formulations using high-performance liquid chromatography (HPLC) technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validated HPLC method for quantifying permethrin in pharmaceutical formulations.
Garcie E, et al.
Journal of Pharmaceutical and Biomedical Analysis, 24(5-6), 999-1004 (2001)
Ludovic P Ahoua Alou et al.
Parasites & vectors, 5, 289-289 (2012-12-13)
The growing development of pyrethroid resistance constitutes a serious threat to malaria control programmes and if measures are not taken in time, resistance may compromise control efforts in the foreseeable future. Prior to Long Lasting Insecticidal Nets (LLINs) distribution in
Mohamad Goldust et al.
Annals of parasitology, 59(1), 31-35 (2013-07-09)
Scabies is a common parasitic infestation that is an important public-health problem in many resource-poor regions. It is commonly treated with the insecticides but the treatment of choice is still controversial. This study aimed at comparing the efficacy of permethrin
Ting Yang et al.
Journal of medical entomology, 50(3), 585-593 (2013-06-28)
Insecticides, especially pyrethroids, are important components in the vector-control effort. To better understand the development of resistance, the current study characterized resistance profiles in individual single-egg-raft colonies of a field population of Culex quinquefasciatus Say, HAmCq(G0). Our study, with 104
Sarah Gray et al.
The New Zealand medical journal, 126(1373), 53-61 (2013-06-26)
To assess the acceptability and feasibility of delivering targeted primary health care in a decile one primary school setting. A pilot public health nurse (PHN)-led clinic was set up in a South Auckland primary school (roll approximately 400). The clinic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service