Skip to Content
Merck
All Photos(1)

Key Documents

28941

Supelco

Mercury Standard for ICP

TraceCERT®, 1 g/L Hg in nitric acid (nominal concentration)

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€114.00

€114.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€114.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€114.00


Estimated to ship on31 May 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

1 g/L Hg in nitric acid (nominal concentration)

technique(s)

ICP: suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

Mercury standard may have been used as reference standard in HPLC-ICP-MS analysis for the determination of methyl and inorganic mercury from biological samples.[1]

Analysis Note

1 g/L Hg in 12% nitric acid, prepared with high purity Hg metal, HNO3 and water

Other Notes

For a complete product listing of our TraceCERT® range of CRMs for ICP and AAS, technical information and example certificates, please visit our ICP & AAS standards website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT RE 2

Target Organs

Kidney

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meng Wang et al.
Talanta, 71(5), 2034-2039 (2007-03-30)
A mild, efficient and convenient extraction method of using 2-mercaptoethanol contained extractant solution combined with an incubator shaker for determination of mercury species in biological samples by HPLC-ICP-MS has been developed. The effects of the concentration of 2-mercaptoethanol, the composition
Nabi Shariatifar et al.
Biological trace element research, 198(2), 721-731 (2020-03-20)
Rice is one of the most valuable nutrients in the diet of most people in the world. The aim of this study was to evaluate the effect of various pre-cooking (washing, soaking) and cooking processes (traditional and rinse) of rice
Preeyanut Butmee et al.
Talanta, 221, 121669-121669 (2020-10-21)
Mercury ion (Hg2+) is considered to be one of the most toxic heavy metal ions and can cause adverse effects on kidney function, the central nervous system, and the immune system. Therefore, it is important to develop a fast and

Articles

Recent advances in analytical tech for heavy metal detection ensure food safety, with products and services supporting trust in consumption.

A 4-step comprehensive ICP-MS workflow including sample homogenization, with standard addition calibration method, to determine heavy metals in Cannabis sativa.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service