Skip to Content
Merck
All Photos(1)

Key Documents

47938

Supelco

JMHW VOC Mix

1000 μg/mL in methanol, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

certified reference material

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 5 × 1 mL

concentration

1000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Application

JMHW VOC Mix has been used as an analytical reference standard for the quantification of volatile organic compounds in residential buildings using gas chromatography coupled to mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Benzene
    Bromodichloromethane
    Bromoform
    Carbon tetrachloride
    Chloroform
    Dibromochloromethane
    1,4-Dichlorobenzene
    1,2-Dichloroethane
    1,1-Dichloroethylene
    cis-1,2-Dichloroethylene
    trans-1,2-Dichloroethylene
    Dichloromethane
    1,2-Dichloropropane
    cis-1,3-Dichloropropene
    trans-1,3-Dichloropropene
    Tetrachloroethylene
    Toluene
    1,1,1-Trichloroethane
    1,1,2-Trichloroethane
    Trichloroethylene
    o-Xylene
    m-Xylene
    p-Xylene
    See All (23)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1A - Flam. Liq. 2 - Muta. 1B - Ozone 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

EU REACH Annex XVII (Restriction List)

EU REACH Annex XIV (Authorisation List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service