Skip to Content
Merck
All Photos(1)

Key Documents

CRM48775

Supelco

EPA VOC Mix 1

certified reference material, 2000 μg/mL each component in methanol, ampule of 1 mL

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€41.30

€41.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
€41.30

About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

€41.30


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    sec-Butylbenzene
    tert-Butylbenzene
    Chlorobenzene
    2-Chlorotoluene
    4-Chlorotoluene
    1,2-Dichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    Propylbenzene
    o-Xylene
    p-Xylene
    See All (11)

Signal Word

Danger

WGK

WGK 3

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service