Skip to Content
Merck
All Photos(1)

Key Documents

47868

Supelco

D-Biotin

analytical standard

Synonym(s):

Biotin, D-Biotin, Bios II, Coenzyme R, Vitamin B7, Vitamin H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O3S
CAS Number:
Molecular Weight:
244.31
Beilstein:
86838
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:

grade

analytical standard

feature

standard type water soluble vitamin

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

231-233 °C (lit.)

application(s)

pharmaceutical (small molecule)
vitamins, nutraceuticals, and natural products

format

neat

storage temp.

2-8°C

SMILES string

[H][C@]12CS[C@@H](CCCCC(O)=O)[C@@]1([H])NC(=O)N2

InChI

1S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7-,9-/m0/s1

InChI key

YBJHBAHKTGYVGT-ZKWXMUAHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-biotin is water soluble vitamin, which belongs to the B-complex group of vitamins. It generally serves as a covalently-bound coenzyme for several carboxylases. It is an essential micronutrient required for normal cell functions and can be obtained both dietary sources as well as gut microbiota.

Application

D-Biotin may be used as an analytical reference standard for the quantification of the analyte in food samples using high-performance liquid chromatography (HPLC)/avidin-binding assay.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmacological Effects of Biotin in Animals
L R-N and C F-M
Mini-Reviews in Medicinal Chemistry, 17, 529-529 (2017)
Determination of the biotin content of select foods using accurate and sensitive HPLC/avidin binding
Staggs GC, et al.
Journal of food composition and analysis, 17(6), 767-776 (2004)
Lipopolysaccharide inhibits colonic biotin uptake via interference with membrane expression of its transporter: a role for a casein kinase 2-mediated pathway
Lakhan R and Said.MS
American Journal of Physiology. Cell Physiology, 312, C376-C384 (2017)
Konstantinos Ampatzis et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(26), 10875-10886 (2013-06-28)
In vertebrates, spinal circuits drive rhythmic firing in motoneurons in the appropriate sequence to produce locomotor movements. These circuits become active early during development and mature gradually to acquire the flexibility necessary to accommodate the increased behavioral repertoire of adult
Tazu Aoki et al.
Neuron, 78(5), 881-894 (2013-05-21)
The encoding of long-term associative memories for learned behaviors is a fundamental brain function. Yet, how behavior is stably consolidated and retrieved in the vertebrate cortex is poorly understood. We trained zebrafish in aversive reinforcement learning and measured calcium signals

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service