Skip to Content
Merck
All Photos(1)

Key Documents

0711610

Sigma-Aldrich

1-N-(4,4′-Dimethoxytrityl)-D-(+)-biotin

>95%

Synonym(s):

D-Biotin 4,4′-dimethoxytrityl ester, DMT-Biotin

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€439.00

€439.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€439.00

About This Item

Empirical Formula (Hill Notation):
C31H34N2O5S
CAS Number:
Molecular Weight:
546.68
Beilstein:
7673243
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.03

€439.00


Check Cart for Availability

Request a Bulk Order

Assay

>95%
98% (HPLC)

contains

≤3% Pyridine as stabilizer

SMILES string

COc1ccc(cc1)C(N2C3CS[C@@H](CCCCC(O)=O)C3NC2=O)(c4ccccc4)c5ccc(OC)cc5
COc1ccc(cc1)C(N2C3CS[C@@H](CCCCC(O)=O)C3NC2=O)(c4ccccc4)c5ccc(OC)cc5

InChI

1S/C31H34N2O5S/c1-37-24-16-12-22(13-17-24)31(21-8-4-3-5-9-21,23-14-18-25(38-2)19-15-23)33-26-20-39-27(29(26)32-30(33)36)10-6-7-11-28(34)35/h3-5,8-9,12-19,26-27,29H,6-7,10-11,20H2,1-2H3,(H,32,36)(H,34,35)/t26-,27-,29-/m0/s1

InChI key

RMWVJZXKPGLZEJ-YCVJPRETSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. N. Kremsky et al.
Tetrahedron Letters, 37, 4313-4313 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service