Skip to Content
Merck
All Photos(1)

Key Documents

46911

Supelco

Deoxynivalenol solution

200 μg/mL in ethyl acetate: methanol (95:5), analytical standard

Synonym(s):

3α,7α,15-Trihydroxy-12,13-epoxytrichothec-9-en-8-one, DON, Vomitoxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20O6
CAS Number:
Molecular Weight:
296.32
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1 mL

concentration

200 μg/mL in ethyl acetate: methanol (95:5)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

single component solution

storage temp.

−20°C

SMILES string

CC1=C[C@H]2O[C@@H]3[C@H](O)C[C@@](C)([C@]34CO4)[C@@]2(CO)[C@H](O)C1=O

InChI

1S/C15H20O6/c1-7-3-9-14(5-16,11(19)10(7)18)13(2)4-8(17)12(21-9)15(13)6-20-15/h3,8-9,11-12,16-17,19H,4-6H2,1-2H3/t8-,9-,11-,12-,13-,14-,15+/m1/s1

InChI key

LINOMUASTDIRTM-QGRHZQQGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

26.6 °F - closed cup

Flash Point(C)

-3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amal Halawa et al.
Archives of animal nutrition, 67(2), 134-146 (2013-03-26)
Deoxynivalenol (DON) is one of the most important trichothecenes, due to its worldwide distribution and common contamination of animal feed. It mainly affects the gastrointestinal tract and the immune system with a high susceptibility for swine. Lipopolysaccharides (LPS) are endotoxins
Sven Dänicke et al.
Toxicology letters, 220(2), 172-180 (2013-04-23)
The systemic effects of the Fusarium toxin deoxynivalenol (DON) and of bacterial lipopolysaccharides (LPS) were studied in male castrated pigs (40.4 ± 3.7 kg) infused intravenously with either DON or LPS alone (100 μg DON/kg/h, 7.5 μg/LPS/kg/h), or together (100
Brenna M Flannery et al.
Toxicology, 304, 192-198 (2013-01-10)
Consumption of the trichothecene deoxynivalenol (DON) suppresses growth in experimental animals - an adverse effect that was used to establish the tolerable daily intake for this toxin. DON ingestion has been recently found to suppress plasma insulin-like growth factor acid-labile
Dirk Rohweder et al.
Archives of animal nutrition, 67(1), 37-47 (2013-01-23)
Fusarium infections do not only affect the grain, but also the rest of the plant, which result in contamination of plants with the mycotoxin deoxynivalenol (DON). The bioavailability of DON may be influenced by the matrix due to the differences
Joelma Lucioli et al.
Toxicon : official journal of the International Society on Toxinology, 66, 31-36 (2013-02-14)
Trichothecenes induce changes in the intestinal barrier function through decreased expression of cell junction proteins and apoptosis of enterocytes. The mitogen activated protein kinases (MAPK) play an important role in the signaling pathways of cell turnover and differentiation. Using ex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service